Skip to Content
Merck

U6753

Sigma-Aldrich

Ursolic acid

≥90%

Synonym(s):

3β-Hydroxy-12-ursen-28-ic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
Beilstein:
2228563
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Pricing and availability is not currently available.

Quality Level

Assay

≥90%

mp

292 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

[H][C@@]12CC[C@]3(C)[C@]([H])(CC=C4[C@]5([H])[C@@H](C)[C@H](C)CC[C@@]5(CC[C@@]34C)C(O)=O)[C@@]1(C)CC[C@H](O)C2(C)C

InChI

1S/C30H48O3/c1-18-10-15-30(25(32)33)17-16-28(6)20(24(30)19(18)2)8-9-22-27(5)13-12-23(31)26(3,4)21(27)11-14-29(22,28)7/h8,18-19,21-24,31H,9-17H2,1-7H3,(H,32,33)/t18-,19+,21+,22-,23+,24+,27+,28-,29-,30+/m1/s1

InChI key

WCGUUGGRBIKTOS-GPOJBZKASA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
8979767231503240595
mp

292 °C (dec.) (lit.)

mp

292 °C (dec.) (lit.)

mp

-

mp

292 °C (dec.) (lit.)

storage temp.

2-8°C

storage temp.

-

storage temp.

2-8°C

storage temp.

-

Gene Information

mouse ... Ifng(15978), Nos2(18126)

Gene Information

-

Gene Information

-

Gene Information

-

General description

Ursolic acid (UA) is a hydroxyl pentacyclic triterpenoic acid (HPTA), which exhibits anti-bacterial,[1] anti-cancer,[2] anti-oxidative and anti-inflammatory effects.[3] It can also promote neuroregeneration after peripheral nerve injury.[3] UA can enhance sleep duration by activating the GABAergic neurotransmitter system.[4] [GABA= γ-aminobutyric acid]

Application

Ursolic acid can be used as a starting material to synthesize ethoxycarbonylmethyl ursolate (ECU) by reacting with ethyl chloroacetate. ECU is further used as a key intermediate in the total synthesis of a biologically important compound 3-acetyl-2-[(un)substituted phenyl]-2,3-dihydro-1,3,4-oxadiazol-5-Me 3-O-acetylursolate.[5] Ursolic acid can also be used to prepare novel heterocyclic derivatives for various activity studies.[6]

Biochem/physiol Actions

Triterpenoid found in a variety of fruits, including apples. Cardioprotective and anti-tumor agent. Under study as a potential Alzheimer′s disease therapeutic due to its inhibitory effect on the interactions between amyloid-β and the CD36 receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

580.5 °F

Flash Point(C)

304.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ursolic acid enhances pentobarbital-induced sleeping behaviors via GABAergic neurotransmission in mice.
Jeon, SJ, et al.
European Journal of Pharmacology, 762, 443-448 (2015)
Ursolic acid protects hippocampal neurons against kainate-induced excitotoxicity in rats.
Shih YH, et al.
Neuroscience Letters, 363(2), 136-140 (2004)
Synthesis and anti-inflammatory activity of 3-acetyl-2-[(un) substituted phenyl]-2, 3-dihydro-1, 3, 4-oxadiazol-5-methyl 3-O-acetylursolate
Ding-Ju Y, et al.
Chinese Journal of Organic Chemistry, 28(6), 1055-1060 (2008)
Ursolic acid induces neural regeneration after sciatic nerve injury.
Liu B, et al.
Neural Regeneration Research, 8(27), 2510-2510 (2013)
Aljawharah AlQathama et al.
Biomolecules, 10(6) (2020-06-18)
We evaluate how 3-acetylation modulates the in vitro activity of ursolic acid in melanoma cells alone or in combination treatments with quercetin. Anti-proliferative studies on A375 cells and adult human dermal fibroblasts included analyses on cell cycle distribution, caspase activity

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service