Skip to Content
Merck

89797

Supelco

Ursolic acid

analytical standard

Synonym(s):

3β-Hydroxy-12-ursen-28-ic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
Beilstein:
2228563
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.24

Pricing and availability is not currently available.

grade

analytical standard

Quality Level

Assay

≥98.5% (HPLC)

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

mp

292 °C (dec.) (lit.)

application(s)

food and beverages

format

neat

SMILES string

[H][C@@]12CC[C@]3(C)[C@]([H])(CC=C4[C@]5([H])[C@@H](C)[C@H](C)CC[C@@]5(CC[C@@]34C)C(O)=O)[C@@]1(C)CC[C@H](O)C2(C)C

InChI

1S/C30H48O3/c1-18-10-15-30(25(32)33)17-16-28(6)20(24(30)19(18)2)8-9-22-27(5)13-12-23(31)26(3,4)21(27)11-14-29(22,28)7/h8,18-19,21-24,31H,9-17H2,1-7H3,(H,32,33)/t18-,19+,21+,22-,23+,24+,27+,28-,29-,30+/m1/s1

InChI key

WCGUUGGRBIKTOS-GPOJBZKASA-N

Looking for similar products? Visit Product Comparison Guide

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
032405951707770U6753
application(s)

food and beverages

application(s)

food and beverages

application(s)

pharmaceutical (small molecule)

application(s)

-

format

neat

format

-

format

neat

format

-

Quality Level

100

Quality Level

-

Quality Level

-

Quality Level

-

grade

analytical standard

grade

primary reference standard

grade

pharmaceutical primary standard

grade

-

assay

≥98.5% (HPLC)

assay

-

assay

-

assay

≥90%

General description

Ursolic acid belongs to the class of triterpenoid compounds that occurs widely in medicinal herbs, food and other plants.[1] It may possess pharmacological properties like hepta-protection, analgesic, cardiotonic, sedative and tonic effects.[1]

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Biochem/physiol Actions

Triterpenoid found in a variety of fruits, including apples. Cardioprotective and anti-tumor agent. Under study as a potential Alzheimer′s disease therapeutic due to its inhibitory effect on the interactions between amyloid-β and the CD36 receptor.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

580.5 °F

Flash Point(C)

304.7 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Pharmacology of oleanolic acid and ursolic acid
Liu J
Journal of Ethnopharmacology, 49, 57-68 (1995)
Huang-Yao Tu et al.
Bioorganic & medicinal chemistry, 17(20), 7265-7274 (2009-09-18)
Twenty-three ursolic acid (1) derivatives 2-24 including nine new 1 derivatives 5, 7-11, 20-22 were synthesized and evaluated for cytotoxicities against NTUB1 cells (human bladder cancer cell line). Compounds 5 and 17 with an isopropyl ester moiety at C-17-COOH and
Jing-Wei Shao et al.
European journal of medicinal chemistry, 46(7), 2652-2661 (2011-04-26)
Twenty-three ursolic acid (1) derivatives 2-24 (ten novel compounds 8-10, 14-17 and 22-24) modified at the C-3 and the C-28 positions were synthesized, and their structures were confirmed by IR, (1)H NMR, MS, and elemental analysis. The single crystals of
Jin-Feng Hu et al.
Journal of natural products, 69(1), 118-120 (2006-01-31)
One new (1) and four known (2-5) ursene triterpenes with potent inhibition of the formation of the bacterial biofilm Pseudomonas aeruginosa PA01 were obtained from Diospyros dendo using a high-throughput natural products chemistry procedure. These compounds were isolated as mass-limited
MinKyun Na et al.
Journal of natural products, 69(11), 1572-1576 (2006-11-28)
Inhibition of protein tyrosine phosphatase-1B (PTP1B) has been proposed as a therapy for treatment of type-2 diabetes and obesity. Bioassay-guided fractionation of an EtOAc-soluble extract of the root bark of Erythrina mildbraedii, using an in vitro PTP1B inhibitory assay, resulted

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service