Skip to Content
Merck
All Photos(1)

Key Documents

855057

Sigma-Aldrich

Betulinic acid

technical grade, 90%

Synonym(s):

3β-Hydroxy-20(29)-lupaene-28-oic acid, Lupatic acid, Mairin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
EC Number:
MDL number:
UNSPSC Code:
85151701
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

90%

form

powder

optical activity

[α]20/D +7.8°, c = 0.9 in pyridine

mp

295-298 °C (dec.) (lit.)

storage temp.

2-8°C

SMILES string

CC(=C)[C@@H]1CC[C@@]2(CC[C@]3(C)[C@H](CC[C@@H]4[C@@]5(C)CC[C@H](O)C(C)(C)[C@@H]5CC[C@@]34C)[C@@H]12)C(O)=O

InChI

1S/C30H48O3/c1-18(2)19-10-15-30(25(32)33)17-16-28(6)20(24(19)30)8-9-22-27(5)13-12-23(31)26(3,4)21(27)11-14-29(22,28)7/h19-24,31H,1,8-17H2,2-7H3,(H,32,33)/t19-,20+,21-,22+,23-,24+,27-,28+,29+,30-/m0/s1

InChI key

QGJZLNKBHJESQX-FZFNOLFKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Betulinic acid is a lupane-type triterpene, which can be prepared from betulin via oxidation using chromium oxide (VI). Along with anti-HIV-1 and anti-tumor activity, it also shows other bioactivities such as anti-inflammatory, antibacterial, in vitro antimalarial, anthelmintic and antioxidant activities.

Biochem/physiol Actions

Betulinic acid, a pentacyclic triterpene, selectively induces apoptosis in tumor cells by directly activating the mitochondrial pathway of apoptosis through a p53- and CD95-independent mechanism.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Li Kong et al.
Antioxidants (Basel, Switzerland), 10(2) (2021-01-28)
T-2 toxin, which is mainly produced by specific strains of Fusarium in nature, can induce immunotoxicity and oxidative stress, resulting in immune organ dysfunction and apoptosis. Betulinic acid (BA), a pentacyclic triterpenoids from nature plants, has been demonstrated to possess
Rafael Alcalá-Vida et al.
EMBO molecular medicine, 13(2), e12105-e12105 (2020-12-29)
Lamins are crucial proteins for nuclear functionality. Here, we provide new evidence showing that increased lamin B1 levels contribute to the pathophysiology of Huntington's disease (HD), a CAG repeat-associated neurodegenerative disorder. Through fluorescence-activated nuclear suspension imaging, we show that nucleus
Sami Alakurtti et al.
Bioorganic & medicinal chemistry, 18(4), 1573-1582 (2010-02-02)
Betulin, a naturally occurring abundant triterpene is converted in four steps to 3,28-di-O-acetyllupa-12,18-diene. When various 4-substituted urazoles were oxidized to the corresponding urazines with iodobenzene diacetate in the presence of 3,28-di-O-acetyllupa-12,18-diene, new heterocyclic betulin derivatives were produced. These betulin derivatives
Rita C Santos et al.
Bioorganic & medicinal chemistry, 17(17), 6241-6250 (2009-08-14)
A series of new imidazole carboxylic esters (carbamates) and N-acylimidazole derivatives of betulin and betulinic acid (14-29) have been synthesized. The new compounds were screened for in vitro cytotoxicity activity against human cancer cell lines HepG2, Jurkat and HeLa. A
Yan-Fang Sun et al.
Food chemistry, 138(2-3), 1998-2007 (2013-02-16)
Betulinic acid (BA), a natural pentacyclic triterpenoid, was isolated from sour jujube fruits for the first time. An inclusion complex comprising BA and β-cyclodextrin (β-CD) was formed to improve the dissolution of BA, but little is known about its anticancer

Articles

Chronic inflammation is an underlying factor in the development and progression of many of the chronic diseases of aging, such as arthritis, atherosclerosis, diabetes, and cancer.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service