Skip to Content
Merck
All Photos(3)

Key Documents

O5504

Sigma-Aldrich

Oleanolic acid

≥97%

Synonym(s):

(+)-Oleanolic acid, 3beta-Hydroxyolean-12-en-28-oic acid, Astrantiagenin C, Caryophyllin, Oleanic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C30H48O3
CAS Number:
Molecular Weight:
456.70
EC Number:
MDL number:
UNSPSC Code:
12352002
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97%

form

powder

color

light yellow

mp

>300 °C (lit.)

storage temp.

2-8°C

SMILES string

[H][C@@]12CC(C)(C)CC[C@@]1(CC[C@]3(C)C2=CC[C@]4([H])[C@@]5(C)CC[C@H](O)C(C)(C)[C@]5([H])CC[C@@]34C)C(O)=O

InChI

1S/C30H48O3/c1-25(2)14-16-30(24(32)33)17-15-28(6)19(20(30)18-25)8-9-22-27(5)12-11-23(31)26(3,4)21(27)10-13-29(22,28)7/h8,20-23,31H,9-18H2,1-7H3,(H,32,33)/t20-,21-,22+,23-,27-,28+,29+,30-/m0/s1

InChI key

MIJYXULNPSFWEK-GTOFXWBISA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

General description

Oleanolic acid is a hydroxyl pentacyclic triterpenoic acid (HPTA), which exhibits antibacterial, antitumor and antileishmanial activity.

Application

Oleanolic acid has been used as a reference compound during its quantification in the roots of Cissampelos pareria Linn.var hirsuta by high performance thin layer chromatography (HPTLC).

Biochem/physiol Actions

Triterpenoid isolated from medicinal plants. Antitumor and hepatoprotective agent. Oleanolic acid has therapeutic potential for neurodegenerative diseases.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oleanolic acid (OA) as an antileishmanial agent: Biological evaluation and in silico mechanistic insights.
Melo TS, et al.
Parasitology International, 65(3), 227-237 (2016)
Apurva Bhave et al.
Journal of food and drug analysis, 25(3), 681-690 (2017-09-16)
In this study, an in-depth analysis of the unique set of rosehip samples from 71 Rosa genotypes was conducted with the aim to identify the most suitable ones for applications in the food and pharmaceutical industries based on the content
Validated HPTLC method for qualitative and quantitative estimation of oleanolic acid in roots of Cissampelos pareira Linn. var. hirsuta, Menispermaceae.
Patel VG, et al.
Der Pharmacia Lettre, 7(5), 300-306 (2015)
Amélia M Silva et al.
Pharmaceutics, 11(8) (2019-07-28)
Oleanolic (OA) and ursolic (UA) acids are recognized triterpenoids with anti-cancer properties, showing cell-specific activity that can be enhanced when loaded into polymeric nanoparticles. The cytotoxic activity of OA and UA was assessed by Alamar Blue assay in three different
Ursolic, oleanolic and betulinic acids: antibacterial spectra and selectivity indexes.
Fontanay S, et al.
Journal of Ethnopharmacology, 120(2), 272-276 (2008)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service