Skip to Content
Merck
All Photos(1)

Key Documents

T67806

Sigma-Aldrich

Triisopropyl phosphite

95%

Synonym(s):

Isopropyl phosphite, Tri-2-propyl phosphite, Triisopropoxyphosphine, Tris(isopropyl) phosphite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CHO]3P
CAS Number:
Molecular Weight:
208.24
Beilstein:
1701528
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<2 mmHg ( 20 °C)

Quality Level

Assay

95%

refractive index

n20/D 1.411 (lit.)

bp

63-64 °C/11 mmHg (lit.)

density

0.844 g/mL at 25 °C (lit.)

SMILES string

CC(C)OP(OC(C)C)OC(C)C

InChI

1S/C9H21O3P/c1-7(2)10-13(11-8(3)4)12-9(5)6/h7-9H,1-6H3

InChI key

SJHCUXCOGGKFAI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Triisopropyl phosphite can be used as a reactant:
  • With Ru-based indenylidene complexes to form 1st generation complexes for metathesis reactions.
  • In Perkow-type reaction to synthesize compounds containing polarized carbon-carbon double bonds.
  • For the synthesis of phosphonohydrazines by reacting with arylamines and isoamyl nitrite.

It can also be used as an alternative to triphenylphosphine in Mitsunobu reaction to facilitate the isolation of products.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral - Skin Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

154.4 °F - closed cup

Flash Point(C)

68 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruthenium indenylidene ?1st generation? olefin metathesis catalysts containing triisopropyl phosphite
Guidone S, et al.
Beilstein Journal of Organic Chemistry, 11, 1520-1520 (2015)
Mitsunobu reactions of nucleoside analogs using triisopropyl phosphite-DIAD
Veliz EA and Beal PA
Tetrahedron Letters, 47(18), 3153-3156 (2006)
Nef-isocyanide-Perkow synthesis of new polarized olefins containing 2-dicyanomethylene-2, 3-dihydrothiazole and 2-dicyanomethylene-1, 3-dithiole moieties
Yavari I, et al.
Tetrahedron, 69(11), 2462-2467 (2013)
Convenient synthesis of phosphonohydrazines from arylamines
Kent GT, et al.
Tetrahedron Letters, 57(19), 2097-2099 (2016)
Su Mei Liu et al.
Marine pollution bulletin, 60(9), 1591-1599 (2010-04-30)
Inorganic or bulk organic chemical indicators, including organic carbon (OC), total nitrogen, organic nitrogen (ON), fixed ammonium (N(fix)), exchangeable ammonium, exchangeable nitrate, organic phosphorus (OP), inorganic phosphorus (IP), and biogenic silica (BSi), were examined in a 3-m core collected in

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service