Skip to Content
Merck
All Photos(1)

Key Documents

441805

Sigma-Aldrich

Tris(nonylphenyl) phosphite

Synonym(s):

Tri(monononylphenyl) phosphite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C9H19C6H4O)3P
CAS Number:
Molecular Weight:
689.00
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

liquid

Quality Level

refractive index

n20/D 1.528 (lit.)

bp

>360 °C (lit.)

density

0.99 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCc1ccccc1OP(Oc2ccccc2CCCCCCCCC)Oc3ccccc3CCCCCCCCC

InChI

1S/C45H69O3P/c1-4-7-10-13-16-19-22-31-40-34-25-28-37-43(40)46-49(47-44-38-29-26-35-41(44)32-23-20-17-14-11-8-5-2)48-45-39-30-27-36-42(45)33-24-21-18-15-12-9-6-3/h25-30,34-39H,4-24,31-33H2,1-3H3

InChI key

WGKLOLBTFWFKOD-UHFFFAOYSA-N

General description

Tris(nonylphenyl) phosphite (TNPP) is an antioxidant that is majorly used as a stabilizer to improve the performance of polyethylene by decomposing the hydroperoxides. It can also be used to enhance the thermal stability by extending the polymeric chains.

Application

TNPP in combination with irganox is used to protect polyamide 6 (PA6) from oxidative degradation during the preparation of flame retardant nanocomposites. TNPP can be used as a stabilizer that prevents the molecular weight reduction and increases the tensile strength of the composite. It can also be used as a chain extender to improve the polymeric viscosity during the melt-blending of poly(hydroxybutyrate-co-hydroxyvalerate) based clay nanocomposites which find potential application as packaging materials.

Signal Word

Danger

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Repr. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

404.6 °F - closed cup

Flash Point(C)

207 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

LC-MS/MS analytical procedure to quantify tris (nonylphenyl) phosphite, as a source of the endocrine disruptors 4-nonylphenols, in food packaging materials
Mottier P, et al.
Food Additives & Contaminants. Part A, Chemistry, Analysis, Control, Exposure & Risk Assessment, 31(5), 962-972 (2014)
On the use of tris (nonylphenyl) phosphite as a chain extender in melt-blended poly (hydroxybutyrate-co-hydroxyvalerate)/clay nanocomposites: Morphology, thermal stability, and mechanical properties
Gonzalez?Ausejo J, et al.
Journal of Applied Polymer Science, 133(2), 118-129 (2016)
Bio-based solution-cast blend films based on polylactic acid and polyhydroxybutyrate: Influence of pyromellitic dianhydride as chain extender on the morphology, dispersibility, and crystallinity
Dawin TP, et al.
Progress in Organic Coatings, 119(5), 23-30 (2018)
Halloysite nanotubes loaded with liquid organophosphate for enhanced flame retardancy and mechanical properties of polyamide 6
Boonkongkaew M and Sirisinha K
Journal of materials science & technology, 119(5), 1-13 (2018)
Effects of chain-extending stabilizer on bioplastic poly (lactic acid)/polyamide blends compatibilized by reactive extrusion
Gug J, et al.
Polymer Degradation and Stability, 153(5), 118-129 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service