Skip to Content
Merck
All Photos(2)

Key Documents

554669

Sigma-Aldrich

Triisopropyl phosphate

95%

Synonym(s):

NSC 46370, NSC 62275, Phosphoric acid triisopropyl ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
PO(OCH(CH3)2)3
CAS Number:
Molecular Weight:
224.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

refractive index

n20/D 1.4070 (lit.)

bp

224 °C (lit.)

density

0.970 g/mL at 25 °C (lit.)

SMILES string

CC(C)OP(=O)(OC(C)C)OC(C)C

InChI

1S/C9H21O4P/c1-7(2)11-14(10,12-8(3)4)13-9(5)6/h7-9H,1-6H3

InChI key

OXFUXNFMHFCELM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Triisopropyl phosphate, an ester of secondary alcohol, is a non-phosphorylating organophosphorus compound. It is formed as a by-product during the synthesis of haloalkylphosphonates. Triisopropyl phosphate can be prepared from isopropyl alcohol by reacting with phosphorus oxychloride. It can also be obtained from triisopropyl phosphite.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Aquatic Acute 1 - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

73.4 °F - closed cup

Flash Point(C)

23 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Efficient and `green?microwave-assisted synthesis of haloalkylphosphonates via the Michaelis?Arbuzov reaction.
Jansa P, et al.
Green Chemistry, 13(4), 882-888 (2011)
Reactions of Trialkyl Phosphates, Alkyl Acetates, and Tertiary Butyl Hypochlorite in the Friedel-Crafts Syntheses1.
Berman N and Lowy A.
Journal of the American Chemical Society, 60(11), 2596-2597 (1938)
Dealkylation and debenzylation of triesters of phosphoric acid. Phosphorylation of hydroxy and amino compounds.
Zervas L and Dilaris I.
Journal of the American Chemical Society, 77(20), 5354-5357 (1955)
Physicochemical characteristics and sorption capacities of heavy metal ions of activated carbons derived by activation with different alkyl phosphate triesters
Wang J, et al.
Applied Surface Science, 316, 443-450 (2014)
Carbonium ion formation in solvolysis of phosphate triesters.
Cox Jr JR and Newton MG.
The Journal of Organic Chemistry, 34(9), 2600-2605 (1969)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service