Skip to Content
Merck
All Photos(2)

Key Documents

934712

Sigma-Aldrich

NMC811

greener alternative

electrode sheet, aluminum substrate, size 5 in. × 10 in.

Synonym(s):

Lithium Nickel Manganese Cobalt Oxide, NMC 811 cathode

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
LiNi0.8Mn0.1Co0.1O2
Molecular Weight:
97.28
UNSPSC Code:
26111700
NACRES:
NA.21

material

aluminum substrate

Quality Level

grade

battery grade

description

Nominal Voltage:3.75 V vs. Li/Li+

form

sheet

composition

Active material loading 10.0 mg/cm2 ± 5%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

size

5 in. × 10 in.

thickness

16 μm , aluminum current collector
60 μm , excluding current collector

particle size

10-13 μm (active material characteristic)

capacity

2.0 mAh/cm2±5 % (Areal capacity)

application(s)

battery manufacturing

greener alternative category

General description

NMC811, electrode sheet, aluminum substrate, is a ready-to-use cathode for lithium-ion battery research. NMC811 is a quaternary lithium metal oxide, with the formula LiNi0.8Mn0.1Co0.1O2, and is a state-of-the-art cathode material for lithium-ion batteries that offers high energy density and cycle lifetimes. The composition of our cathode film is 90% active material, 5% PVDF binder, 5% carbon black.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

The main application of our NMC811 electrode sheet is as a cathode for next-generation lithium-ion batteries (LIBs). The 811 refers to the ratio of metals in the active material that combine to give the high performance: nickel provides high energy density while the manganese and cobalt help to stabilize the spinel crystal structure to extend the cycle lifetime at moderate-high operating temperatures. NMC811 (LiNi0.8Mn0.1Co0.1O2) is a highly nickel-rich NMC composition that is currently attracting considerable attention as a cathode material because it can deliver a high specific capacity of ~200 mAh g–1 with a high average discharge potential of ~3.75 V vs Li/Li+, resulting in a substantial increase of ~25% in energy density compared to LiCoO2.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Evolution of Structure and Lithium Dynamics in LiNi0.8Mn0.1Co0.1O2 (NMC811) Cathodes during Electrochemical Cycling
Marker K, et al.
Chemistry of Materials, 31, 2545?2554-2545?2554 (2019)
Seong-Min Bak et al.
ACS applied materials & interfaces, 6(24), 22594-22601 (2014-11-25)
Thermal stability of charged LiNixMnyCozO2 (NMC, with x + y + z = 1, x:y:z = 4:3:3 (NMC433), 5:3:2 (NMC532), 6:2:2 (NMC622), and 8:1:1 (NMC811)) cathode materials is systematically studied using combined in situ time-resolved X-ray diffraction and mass spectroscopy
Peiyu Hou et al.
Small (Weinheim an der Bergstrasse, Germany), 13(45), 1701802-1701802 (2017-10-05)
The urgent prerequisites of high energy-density and superior electrochemical properties have been the main inspiration for the advancement of cathode materials in lithium-ion batteries (LIBs) in the last two decades. Nickel-rich layered transition-metal oxides with large reversible capacity as well

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service