Skip to Content
Merck
All Photos(2)

Key Documents

765201

Sigma-Aldrich

Lithium manganese oxide

greener alternative

spinel, electrode sheet, size 5 in. × 10 in.

Synonym(s):

LMO, Lithium manganese(III,IV) oxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiMn2O4
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

description

Nominal Voltage: 4.7 V, Li/Li+

Assay

≥98%

form

sheet

composition

loading, ≥80%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

≥80% loading

size

5 in. × 10 in.

thickness

~25 μm

particle size

10-13 μm (typical)

capacity

110 mAh/g
120 mAh/g(nominal at 0.1C)

mp

>1000 °C

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[O-][Mn]=O.O=[Mn]=O

InChI

1S/Li.2Mn.4O/q+1;;;;;;-1

InChI key

VLXXBCXTUVRROQ-UHFFFAOYSA-N

General description

Lithium manganese oxide (LMO) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

LMO spinel electrode sheet is a cost-effective and thermally stable cathode material. It has a three-dimensional framework that is useful in the fabrication of lithium-ion batteries for hybrid electrical vehicles (HEVs).

Other Notes

Crystal Structure: Spinel, cubic, Fd-3m

Operating Condiditons:
  • Recommended maximum charge voltage: 5.0 V vs Li/Li+
  • Recommended maximum charge current: 3 C
  • Recommended cut-off voltage for discharge: 3.5 V vs Li/Li+
  • Recommended maximum discharge current: 5 C

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guyomard, D.; et al.
Journal of the Electrochemical Society, 139, 937-937 (1992)
Kim, D.; et al.
Journal of the Electrochemical Society, 159, A193-A193 (2012)
Lattice vibrations of materials for lithium rechargeable batteries I. Lithium manganese oxide spinel
Julien CM and Massot M
Materials Science and Engineering, B, 97(3), 217-230 (2003)
Improved lithium manganese oxide spinel/graphite Li-ion cells for high-power applications
Amine K, et al.
Journal of Power Sources, 129(1), 14-19 (2004)
Preparation of spinel lithium manganese oxide by aqueous co-precipitation
Naghash AR and Lee JY
Journal of Power Sources, 85(2), 284-293 (2000)

Articles

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service