Skip to Content
Merck
All Photos(1)

Key Documents

932558

Sigma-Aldrich

Lithium Nickel Manganese Cobalt Oxide (NMC111) Crosslinked Polyaniline Composite

Cathode for Lithium ion Battery

Synonym(s):

Doped polyaniline, Lithium Nickel Manganese Cobalt Oxide encapsulated in 3D crosslinked polyaniline, Lithium Nickel Manganese Cobalt Oxide particle, NMC111 in doped polyaniline matrix, NMC111/PANI composite

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

Quality Level

form

solid

application(s)

battery manufacturing

General description

Lithium nickel manganese cobalt oxide (NMC111) material is among the most promising cathode materials for Li-ion batteries (LIBs) because of its high energy density. Doped polyaniline (PANI) is an electronically conductive, semi-flexible polymer, used in composites to enhance conductivity, processability, and thermal stability. This NMC composite product consists of NMC111 particles (7.5±2.5µm D50) encapsulated in a 3D cross-linked conductive polyaniline framework with a controlled morphology to promote electronic conductivity and a stabilized solid electrolyte interphase (SEI).

Application

This NMC composite serves as an electrochemically active cathode material for LIBs. The composite not only allows fast electrical and ionic transport, but also help to stabilize the SEI layer, facilitating increased power density and improved cycle-life performance of NMC cathode at high temperature and high voltage.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nanostructured functional hydrogels as an emerging platform for advanced energy technologies
Zhao F., et al.
Advanced Materials, 30, 1801796-1801796 (2018)
Nanostructured conductive polymer gels as a general framework material to improve electrochemical performance of cathode materials in Li-Ion batteries
Shi Y., et al.
Nano Letters, 17, 1906?1914-1906?1914 (2017)
A tunable 3D nanostructured conductive gel framework electrode for high-performance Lithium Ion batteries
Shi Y., et al.
Advanced Materials, 29, 1603922-1603922 (2017)
Review on the synthesis of LiNixMnyCo1-x-yO2 (NMC) cathodes for lithium-ion batteries
Malik, et al.
Materials Today, 28, 101066-101066 (2022)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service