Skip to Content
Merck
All Photos(2)

Key Documents

765163

Sigma-Aldrich

Lithium nickel manganese cobalt oxide

greener alternative

electrode sheet, aluminum substrate, size 5 in. × 10 in.

Synonym(s):

NMC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiNi0.33Mn0.33Co0.33O2
UNSPSC Code:
26111700
NACRES:
NA.23

grade

battery grade

Quality Level

description

Nominal Voltage: 3.5 V, Li/Li+

composition

loading, ≥80%

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

extent of labeling

≥80% loading

size

5 in. × 10 in.

thickness

16 μm

particle size

8-12 μm (typical)

capacity

150 mAh/g(minimum)

mp

>1000 °C

application(s)

battery manufacturing

greener alternative category

General description

Lithium nickel manganese cobalt oxide (NMC) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

NMC electrode sheet has a layered structure and can be used as an active cathode material. It can further be used in combination with single electrolyte interphase (SEI) for the fabrication of lithium-ion batteries.

Other Notes

Crystal Structure: Rhombohedral

Operating Conditions:
  • Recommended maximum charge voltage: 4.4 V vs Li/Li+
  • Recommended maximum charge current: 1C
  • Recommended cut-off voltage for discharge: 2.7 V vs Li/Li+
  • Recommended maximum discharge current: 1 C
  • Recommended charge method: constant current - constant voltage

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Carc. 2 - Skin Sens. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Belharouak, I.; et al.
Journal of Power Sources, 123, 247-247 (2003)
A bi-functional lithium difluoro (oxalato) borate additive for lithium cobalt oxide/lithium nickel manganese cobalt oxide cathodes and silicon/graphite anodes in lithium-ion batteries at elevated temperatures
Lee SJ, et al.
Electrochimica Acta, 137, 1-8 (2014)
Impedance change and capacity fade of lithium nickel manganese cobalt oxide-based batteries during calendar aging
Schmitt J, et al.
Journal of Power Sources, 353, 183-194 (2017)
Choi, J.; Manthiram, A.; J.
Journal of the Electrochemical Society, 152, A1714-A1714 (2005)
Challenges for rechargeable Li batteries
Goodenough JB and Kim Y
Chemistry of Materials, 22(3), 587-603 (2009)

Articles

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

Professor Qiao's review explores stable microstructures for lithium metal fluoride batteries, advancing energy storage technologies.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service