Skip to Content
Merck
All Photos(1)

Key Documents

930156

Sigma-Aldrich

Lithium hexafluorophosphate solution

in ethylene carbonate and ethyl methyl carbonate, 1 M LiPF6 in EC/EMC=30/70 (w/w), battery grade

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
26111700
NACRES:
NA.21

grade

battery grade

Quality Level

Assay

≤99.95% trace metals basis

Related Categories

Application

Lithium hexafluorophosphate solution in ethylene carbonate and dimethyl carbonate is a class of electrolytic solution material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT RE 1 Inhalation - STOT RE 2 Oral

Target Organs

Bone,Teeth, Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Challenges for Rechargeable Li Batteries.
Goodenough J B, et al.
Chemistry of Materials, 22, 587-603 (2010)
Electrochemical investigation of carbonate-based electrolytes for high voltage lithium-ion cells.
Hu Libo, et al.
Journal of Power Sources, 236, 175-180 (2013)
Challenges for Rechargeable Li Batteries.
Goodenough J B, et al.
Chemistry of Materials, 22, 587-603 (2010)
Nonaqueous liquid electrolytes for lithium-based rechargeable batteries.
Kang Xu
Chemical reviews, 104(10), 4303-4417 (2005-01-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service