Skip to Content
Merck
All Photos(2)

Key Documents

746762

Sigma-Aldrich

Lithium hexafluorophosphate solution

greener alternative

in ethyl methyl carbonate, 1.0 M LiPF6 in EMC, battery grade

Synonym(s):

1.0 M LiPF6 EMC, 1.0 M LiPF6 MEC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiPF6
MDL number:
UNSPSC Code:
26111700
PubChem Substance ID:
NACRES:
NA.23

grade

battery grade

Quality Level

form

solution

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

(1.0 M LiPF6 in EMC)

impurities

<15 ppm H2O
<50 ppm HF

color

APHA: <50

bp

100 °C

density

1.12 g/mL at 25 °C (lit.)

anion traces

chloride (Cl-): ≤1 ppm
sulfate (SO42-): ≤2 ppm

cation traces

Ca: ≤1 ppm
Fe: ≤1 ppm
K: ≤1 ppm
Na: ≤1 ppm
Pb: ≤1 ppm

application(s)

battery manufacturing

greener alternative category

SMILES string

F[P-](F)(F)(F)(F)F.[Li+]

InChI

1S/F6P.Li/c1-7(2,3,4,5)6;/q-1;+1

InChI key

AXPLOJNSKRXQPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium hexafluorophosphate solution in ethyl methyl carbonate is a class of electrolytic solution that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

LiPF6 EMC is widely used as an electrolyte that is thermally stable in solvents. It can be mainly used in the fabrication of lithium-ion batteries.
Liquid electrolyte solutions play a key role in lithium ion batteries (LIB) acting as carrier of lithium ions between the cathode and anode. High purity and battery grade electrolyte solutions are thus crucial for lithium ion battery performance. The most common LIB electrolytes are derived from solutions of lithium salt, such as LiPF6 in non-aqueous solvents, example alkyl carbonates or solvent blend. The choice of the electrolyte solution is dependent on both the operating conditions like temperature and the nature of the electrode material in the LIB. The performance of the electrolyte solutions can be further modified with appropriate additives.

The ready-to-use electrolyte solutions are available in different solvent blends and can support a wide variety of lithium ion battery applications. These solutions are high purity and battery grade thus making them also suitable as standards in LIB research. Customized formulations can be made by inter-mixing the electrolyte solutions or by mixing appropriate of additives.

Other Notes

Handling instructions:
  • Do not use with glass equipment
  • All work should be done very quickly under dry air to prevent electrolytes from water uptake and solvent vaporization.

Legal Information

Product of MU Ionic Solutions Corp

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT RE 1 Inhalation

Target Organs

Bone,Teeth

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F

Flash Point(C)

25 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lucht, B. L.;
Energy Production and Storage, 333-333 (2010)
Thermal stability of LiPF6 salt and Li-ion battery electrolytes containing LiPF6
Yang H, et al.
Journal of Power Sources, 161(1), 573-579 (2006)
A low-temperature electrolyte for lithium and lithium-ion batteries
Plichta EJ and Behl WK
Journal of Power Sources, 88(2), 192-196 (2000)
Preparation and characterization of lithium hexafluorophosphate for lithium-ion battery electrolyte
Liu J, et al.
Transactions of Nonferrous Metals Society of China, 20(2), 344-348 (2010)
Scrosati, B.;
Energy & Environmental Science, 4(9), 3287-3287 (2011)

Articles

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

Solid-state Li batteries: Review of solid electrolytes, ion conduction, structures, and electrochemical processes.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service