Skip to Content
Merck
All Photos(1)

Key Documents

930164

Sigma-Aldrich

Lithium hexafluorophosphate solution

in ethylene carbonate and ethyl methyl carbonate, 1.2 M LiPF6 in EC/EMC=30/70 (w/w), battery grade

Synonym(s):

1.2 M LiPF6 in EC/EMC=30/70 (w/w)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiPF6
MDL number:
UNSPSC Code:
26111700
NACRES:
NA.21

grade

battery grade

Quality Level

SMILES string

F[P-](F)(F)(F)(F)F.[Li+]

InChI

1S/F6P.Li/c1-7(2,3,4,5)6;/q-1;+1

InChI key

AXPLOJNSKRXQPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Lithium hexafluorophosphate solution in ethylene carbonate and ethyl methyl carbonate is a class of electrolytic solution that can be used in lithium-ion batteries. A high demand for Lithium-Ion Battery with higher energy, powder density, and density requires a new electrolyte formulation to enhance the properties. This product is the “2nd generation electrolyte”, which is a naming system used by the United States Department of Energy (DOE) in the Advanced Technology Development Program for advanced lithium-ion battery systems. These materials enable the formation of greener and sustainable batteries for electrical energy storage.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1A - STOT RE 1 Inhalation - STOT RE 2 Oral

Target Organs

Bone,Teeth, Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

80.6 °F

Flash Point(C)

27.0 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Challenges for Rechargeable Li Batteries.
Goodenough J B, et al.
Chemistry of Materials, 22, 587-603 (2010)
Fluorinated electrolytes for Li-ion battery: An FEC-based electrolyte for high voltage LiNi0.5Mn1.5O4/graphite couple.
Hu Libo, et al.
Electrochemical Communications, 35, 76-79 (2013)
Electrochemical investigation of carbonate-based electrolytes for high voltage lithium-ion cells.
Hu Libo, et al.
Journal of Power Sources, 236, 175-180 (2013)
Fluorinated electrolytes for 5 V lithium-ion battery chemistry.
Zhang Z, et al.
Energy & Environmental Science, 6, 1806-1810 (2013)
Nonaqueous liquid electrolytes for lithium-based rechargeable batteries.
Kang Xu
Chemical reviews, 104(10), 4303-4417 (2005-01-27)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service