Skip to Content
Merck
All Photos(2)

Key Documents

934828

Sigma-Aldrich

Lithium hexafluorophosphate solution

greener alternative

in ethylene carbonate and ethyl methyl carbonate with vinylene carbonate additive, 1.0 M LiPF6 in EC/EMC=30/70 (w/w) + 1 wt.% VC, battery grade

Synonym(s):

1.0 M LiPF6 in EC/EMC=30/70 (w/w) + 1 wt.% VC

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiPF6
MDL number:
UNSPSC Code:
12352600
NACRES:
NA.21

grade

battery grade

Quality Level

description

Application: Battery Manufacturing

form

(clear liquid)

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

1.0 M (LiPF6)

impurities

≤100.0 ppm H2O
≤250.0 ppm KF

color

colorless to faint yellow

density

1.203 g/mL

application(s)

battery manufacturing

greener alternative category

SMILES string

F[P-](F)(F)(F)(F)F.[Li+]

InChI

1S/F6P.Li/c1-7(2,3,4,5)6;/q-1;+1

InChI key

AXPLOJNSKRXQPA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Our 1.0 M LiPF6 in EC/EMC=30/70 (w/w) with 1 wt.% VC is a battery-grade electrolyte designed for use in lithium-ion batteries. This electrolyte contains a 1.0 M concentration of lithium hexafluorophosphate in a mixture of ethylene carbonate and ethyl methyl carbonate with a vinylene carbonate additive. Our electrolyte is highly pure with carefully minimized impurities of water, acid, and metals. It is safely packaged in an aluminum bottle under inert, dry conditions.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more information.

Application

Our battery-grade 1.0 M LiPF6 in EC/EMC=30/70 (w/w) with 1 wt.% VC electrolyte is optimized for safety, thermal stability, and performance for lithium-ion batteries. The 30/70 blend of EC/EMC combines the high dielectric constant of ethylene carbonate and the low viscosity of ethyl methyl carbonate to afford an electrolyte with a wide electrochemical stability window and excellent wetting. The vinylene carbonate additive helps to form a passivating solid-electrolyte interface (SEI), which improves the electrochemical performance at higher working temperature and resists thermal damage Our electrolyte is compatible with a wide range of cathode and anode materials, including graphite, lithium-metal, LFP, and NMCs making it ideal for use in many battery applications

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1A - Skin Sens. 1 - STOT RE 1 Inhalation - STOT RE 2 Oral

Target Organs

Bone,Teeth, Kidney

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

69.8 °F

Flash Point(C)

21 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Feng-Ni Jiang et al.
Advanced materials (Deerfield Beach, Fla.), 35(12), e2209114-e2209114 (2023-01-08)
Exploring advanced strategies in alleviating the thermal runaway of lithium-metal batteries (LMBs) is critically essential. Herein, a novel electrolyte system with thermoresponsive characteristics is designed to largely enhance the thermal safety of 1.0 Ah LMBs. Specifically, vinyl carbonate (VC) with
Si Yeol Lee et al.
ACS omega, 5(7), 3579-3587 (2020-03-03)
Nanolithia-based materials are promising lithium-ion battery cathodes owing to their high capacity, low overpotential, and stable cyclic performance. Their properties are highly dependent on the structure and composition of the catalysts, which play a role in activating the lithia to
Yanting Jin et al.
Journal of the American Chemical Society, 140(31), 9854-9867 (2018-07-07)
Fluoroethylene carbonate (FEC) and vinylene carbonate (VC) are widely used as electrolyte additives in lithium ion batteries. Here we analyze the solid electrolyte interphase (SEI) formed on binder-free silicon nanowire (SiNW) electrodes in pure FEC or VC electrolytes containing 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service