Skip to Content
Merck
All Photos(3)

Key Documents

913456

Sigma-Aldrich

(R)-AntPhos

≥97%

Synonym(s):

(R)-4-(Anthracen-9-yl)-3-(tert-butyl)-2,3-dihydrobenzo[d][1,3]oxaphosphole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H23O3P
CAS Number:
Molecular Weight:
402.42
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.22

Quality Level

Assay

≥97%

form

powder

optical purity

ee: ≥99% (HPLC)

reaction suitability

reagent type: ligand

functional group

phosphine

SMILES string

P1(COc2c1c(ccc2)c3c4c(cc5c3cccc5)cccc4)C(C)(C)C

InChI

1S/C25H23OP/c1-25(2,3)27-16-26-22-14-8-13-21(24(22)27)23-19-11-6-4-9-17(19)15-18-10-5-7-12-20(18)23/h4-15H,16H2,1-3H3

InChI key

HKAWVLHXUZNLPP-UHFFFAOYSA-N

Related Categories

Application

(R)-AntPhos is a P-chiral monophosphorus ligand used for the asymmetric Suzuki-Miyaura and Miyaura borylation reactions. This ligand is uniquely effective for sterically hindered cross-coupling reactions.

Legal Information

Sold in collaboration with Zejun Pharmaceuticals

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ruofei Cheng et al.
Journal of the American Chemical Society, 140(13), 4508-4511 (2018-03-27)
Carborane cage chirality is an outstanding issue of great interest as the icosahedral carboranes have wide applications in medicinal and materials chemistry. The synthesis of optically active carborane derivatives, whose chirality is associated with the substitution patterns on the polyhedron
Naifu Hu et al.
Angewandte Chemie (International ed. in English), 55(16), 5044-5048 (2016-03-19)
A highly enantioselective alkene aryloxyarylation led to the high-yielding formation of a series of 1,4-benzodioxanes, 1,4-benzooxazines, and chromans containing quaternary stereocenters with excellent enantioselectivity. The sterically bulky and conformationally well defined chiral monophosphorus ligand L4 or L5 was responsible for
Naifu Hu et al.
Journal of the American Chemical Society, 137(21), 6746-6749 (2015-05-06)
The rhodium-catalyzed asymmetric hydroboration of α-arylenamides with BI-DIME as the chiral ligand and (Bpin)2 as the reagent yields for the first time a series of α-amino tertiary boronic esters in good yields and excellent enantioselectivities (up to 99% ee).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service