912964
(R,R)-BABIBOP
≥97%
Synonym(s):
(3R,3′R)-3,3′-Di-tert-butyl-2,2′,3,3′-tetrahydro-4,4′-bibenzo[d][1,3]oxaphosphole
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Empirical Formula (Hill Notation):
C22H28O2P2
CAS Number:
Molecular Weight:
386.40
UNSPSC Code:
12352200
Recommended Products
Assay
≥97%
form
powder
optical purity
ee: ≥99% (HPLC)
reaction suitability
reagent type: ligand
functional group
phosphine
Application
(R,R)-BABIBOP is a P-chiral biaryl biphosphorus ligand used in Pd, Cu, and Rh-catalyzed asymmetric hydrogenations.
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)
Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)
Other Notes
Legal Information
Sold in collaboration with Zejun Pharmaceuticals
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Efficient P-Chiral Biaryl Bisphosphorus Ligands for Palladium-Catalyzed Asymmetric Hydrogenation
Jiang W, et al.
Chin. J. Chem. (2017)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service