764647
Polyethylenimine hydrochloride
linear, average Mn 10,000, PDI ≤1.5
Synonym(s):
Linear PEI HCl salt, Poly(ethylene imine hydrochloride), Poly(ethylene imine) HCl salt
About This Item
Recommended Products
Quality Level
form
solid
mol wt
average Mn 10,000
mp
276-280 °C (decomposition)
PDI
≤1.5
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
This review will focus on the sub-sector of drug delivery concerning the delivery of nucleic acids, with a particular focus on poly(ethyleneimine).
Gene therapy has become one of the most discussed techniques in biomedical research in recent years.
CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.
CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service