Skip to Content
Merck
All Photos(2)

Key Documents

530573

Sigma-Aldrich

Polypyrrole

doped, conductivity 30 S/cm (bulk), extent of labeling: 20 wt. % loading, composite with carbon black

Synonym(s):

PPy

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

form

solid

contains

proprietary organic sulfonic acid as dopant

extent of labeling

20 wt. % loading

mp

>300 °C

solubility

H2O: insoluble
organic solvents: insoluble

InChI

1S/C4H5N/c1-2-4-5-3-1/h1-5H

InChI key

KAESVJOAVNADME-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Polypyrrole (PPy) is a conductive and highly stable polymer. It may be prepared by a standard electrochemical technique. PPy may also be prepared by reacting β-napthalene sulfonic acid (NSA) and ammonium peroxo-disulphate in aqueous medium. The charges on the surfaces can be easily modified by doping the polymer during its synthesis.Solubility and conductivity measurements of PPy doped with camphor sulfonic and dodecyl benzene sulfonic acid has been reported. Electrosensitivity and lower oxidation potential of PPy make it potentially useful for drug delivery, chemical sensors, batteries, ion selective electrodes, biosensor and biochemistry research. 5
Polypyrrole (PPy) is a widely used heterocyclic polymer that is highly conductive and is majorly used in electrochemical applications. It is a cost efficient conducting polymer that has a stable oxidation and high solubility in water. It has a high degree of flexibility than polyaniline and has a density that facilitates a higher capacitance per unit volume.

Application

Inherently conducting polymer based additive. Loading of polypyrrole in carbon black typically 20%. Stable up to at least 300°C in air.
PPy is mainly used in the fabrication of a variety of electrochemical devices which include supercapacitors, chemical sensors, dye sensitized solar cells and lithium-ion batteries.
Conducting polymer.
Conductive additive for thermoplasitics and thermosets.

Packaging

Packaged in glass bottles

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemistry of nucleic acids and proteins, 1(1), 596-612 (2005)
Conducting polymers with superhydrophobic effects as anticorrosion coating
Intelligent Coatings for Corrosion Control edited by Atul Tiwari, Lloyd Hihara, James Rawlins, 409-430 (2014)
Conducting polymers with superhydrophobic effects as anticorrosion coating
Intelligent Coatings for Corrosion Control, 409-430 (2014)
Construction of high-capacitance 3D CoO@ polypyrrole nanowire array electrode for aqueous asymmetric supercapacitor
Zhou C, et al.
Nano Letters, 13(5), 2078-2085 (2013)
Gusphyl A Justin et al.
Conference proceedings : ... Annual International Conference of the IEEE Engineering in Medicine and Biology Society. IEEE Engineering in Medicine and Biology Society. Annual Conference, 2012, 1206-1209 (2013-02-01)
An electronically-controlled drug delivery system (eDDS) for the on-demand release of anti-inflammatory, anti-microbial and analgesic agents to aid in wound healing is currently under development. The loading of several drugs into conductive polymer films and their subsequent on-demand, controlled release

Articles

The application of conducting polymers at the interface with biology is an exciting new trend in organic electronics research.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

Self-healing soft electronic materials offer potential cost savings and reduced electronic waste.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service