Skip to Content
Merck
All Photos(1)

Key Documents

764965

Sigma-Aldrich

Polyethylenimine hydrochloride

linear, average Mn 15,000, PDI <1.4

Synonym(s):

Linear PEI HCl salt, Poly(ethylene imine hydrochloride), Poly(ethylene imine) HCl salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(NHC2H4)nOH·xHCl
UNSPSC Code:
12162002
NACRES:
NA.23

form

solid

mol wt

average Mn 15,000

mp

276-281 °C (decomposition)

PDI

<1.4

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

This review will focus on the sub-sector of drug delivery concerning the delivery of nucleic acids, with a particular focus on poly(ethyleneimine).

Gene therapy has become one of the most discussed techniques in biomedical research in recent years.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

CRISPR/Cas9 delivery via nonviral nanoparticles shows promising advancements for gene editing in disease treatment.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service