Skip to Content
Merck
All Photos(1)

Key Documents

74398

Sigma-Aldrich

Nonylamine

≥99.5% (GC)

Synonym(s):

1-Aminononane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)8NH2
CAS Number:
Molecular Weight:
143.27
Beilstein:
1733633
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.5% (GC)

form

liquid

impurities

≤0.5% water

refractive index

n20/D 1.433 (lit.)
n20/D 1.433

bp

201 °C (lit.)

density

0.782 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCCCCCCN

InChI

1S/C9H21N/c1-2-3-4-5-6-7-8-9-10/h2-10H2,1H3

InChI key

FJDUDHYHRVPMJZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Nonylamine is an alkylamine generally used to insert C9 chains to substrates. It is also used as intercalating material in the exfoliation of titanosilicates and γ-layered zirconium phosphate.

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

165.2 °F - closed cup

Flash Point(C)

74 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inhibition of quorum sensing in gram-negative bacteria by alkylamine-modified cyclodextrins.
Morohoshi, Tomohiro et al.
Journal of Bioscience and Bioengineering, 116(2), 175-179 (2013)
Tetrahydrofuran-based amino acids as library scaffolds.
Edwards, Alison A et al.
Journal of Combinatorial Chemistry, 6(2), 230-238 (2004)
Exfoliated Titanosilicate Material UZAR-S1 Obtained from JDF-L1.
Rubio, C
European Journal of Inorganic Chemistry, 2010(1), 159-163 (2010)
Intercalation Processes of n-Alkyl Monoamines in γ-Zirconium Phosphate.
Alberti, Giulio et al.
Langmuir, 16(9), 4165-4170 (2000)
Detection of Hb-Papio B, a silent mutation of the baboon beta chain, by high performance liquid chromatography. Improved procedures for the separation of globin chains by HPLC.
J B Shelton et al.
Hemoglobin, 6(5), 451-464 (1982-01-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service