Skip to Content
Merck
All Photos(1)

Key Documents

30692

Sigma-Aldrich

Decylamine

≥99.0% (GC)

Synonym(s):

1-Aminodecane, NDA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)9NH2
CAS Number:
Molecular Weight:
157.30
Beilstein:
1735220
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0% (GC)

form

liquid

impurities

≤0.5% water

refractive index

n20/D 1.436 (lit.)

bp

216-218 °C (lit.)

mp

12-14 °C (lit.)

density

0.787 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCCCCCCCCCN

InChI

1S/C10H23N/c1-2-3-4-5-6-7-8-9-10-11/h2-11H2,1H3

InChI key

MHZGKXUYDGKKIU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Application

Decylamine was used in the synthesis of dynole 2-24 series by undergoing reductive amination with 1-(3-(dimethylamino)propyl)-1H-indole-3-carbaldehyde. It was also used as a coordinating solvent to thermolyse the lead piperidine and lead tetrahydroquinoline dithiocarbamate (DTC) complexes.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Linda D Nyamen et al.
Dalton transactions (Cambridge, England : 2003), 41(27), 8297-8302 (2012-05-30)
We report the synthesis of lead piperidine and lead tetrahydroquinoline dithiocarbamate (DTC) complexes and their use as single source precursors for the preparation of anisotropic PbS nanoparticles. The complexes were thermolysed in coordinating solvents such hexadecylamime (HDA), tri-n-octylphosphine oxide (TOPO)
Aneta D Petelska et al.
Colloids and surfaces. B, Biointerfaces, 82(2), 340-344 (2010-10-05)
Monolayers of phosphatidylcholine, fatty acid and amine and binary mixtures phosphatidylcholine-fatty acid or phosphatidylcholine-amine were investigated at the air/water interface. Phosphatidylcholine (lecithin, PC), stearic acid (SA), palmitic acid (PA), decanoic acid (DA) and decylamine (DE) were used to the experiment.
E R Christensen et al.
Environmental toxicology and chemistry, 20(10), 2361-2369 (2001-10-13)
The joint toxicity of nonylamine and decylamine and of atrazine and decylamine was evaluated in assays with the green alga Selenastrum capricornutum based on an isobologram method. In this method, curves of constant response, isoboles, are plotted versus concentrations of
M Tenne et al.
Journal of neurochemistry, 44(5), 1373-1377 (1985-05-01)
Deamination of n-octylamine and n-decylamine has been studied in various tissues using a new bioluminescence technique. Selectivity of n-octylamine and n-decylamine as substrates for monoamine oxidase (MAO) A or B has been determined using both clorgyline and (-)-deprenyl inhibition curves
Rebecca A Bader et al.
Biomacromolecules, 12(2), 314-320 (2011-01-12)
Despite improvements relative to unmodified counterparts, poly(ethylene glycol) (PEG) conjugation may not be the ideal solution for improving circulatory stability of current nanoparticle carriers or free drugs. Polysialic acid (PSA), a natural polymer for which the body possesses no receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service