Skip to Content
Merck
All Photos(2)

Key Documents

T10200

Sigma-Aldrich

Tetradecylamine

95%

Synonym(s):

1-Aminotetradecane, Myristylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)13NH2
CAS Number:
Molecular Weight:
213.40
Beilstein:
1633740
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

bp

162 °C/15 mmHg (lit.)

mp

38-40 °C (lit.)

SMILES string

CCCCCCCCCCCCCCN

InChI

1S/C14H31N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15/h2-15H2,1H3

InChI key

PLZVEHJLHYMBBY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Roza Bouchal et al.
International journal of molecular sciences, 17(2), 223-223 (2016-02-11)
Surface activity and micelle formation of alkylguanidinium chlorides containing 10, 12, 14 and 16 carbon atoms in the hydrophobic tail were studied by combining conductivity and surface tension measurements with isothermal titration calorimetry. The purity of the resulting surfactants, their
Dejian Yu et al.
Nature communications, 11(1), 3395-3395 (2020-07-09)
Ensuring nuclear safety has become of great significance as nuclear power is playing an increasingly important role in supplying worldwide electricity. β-ray monitoring is a crucial method, but commercial organic scintillators for β-ray detection suffer from high temperature failure and
Liandong Feng et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 23(39), 9397-9406 (2017-05-11)
Dual stimuli-responsive nanoparticles capable of fine-tuning drug release to augment therapeutic efficacy have become a promising tool for anticancer drug delivery. However, the rational design of these "smart" nanoparticles for a selective delivery and controlled release of multidrug combinations in
Elnaz Nakhaei et al.
Analytical sciences : the international journal of the Japan Society for Analytical Chemistry, 34(4), 501-504 (2018-04-13)
Therapeutic peptides and diagnostic agents with their molecular size below the renal clearance threshold suffer from short blood circulation time. Here, we report a novel design of peptide-based ligand with a strong binding affinity to human serum albumin (HSA), which
Lourdes Pérez et al.
International journal of molecular sciences, 21(23) (2020-12-02)
The surface activity, aggregates morphology, size and charge characteristics of binary catanionic mixtures containing a cationic amino acid-derived surfactant N(π), N(τ)-bis(methyl)-L-Histidine tetradecyl amide (DMHNHC14) and an anionic surfactant (the lysine-based surfactant Nα-lauroyl-Nεacetyl lysine (C12C3L) or sodium myristate) were investigated for

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service