Skip to Content
Merck
All Photos(1)

Key Documents

445312

Sigma-Aldrich

Hexadecylamine

98%

Synonym(s):

1-Aminohexadecane, Cetylamine, HAD

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)15NH2
CAS Number:
Molecular Weight:
241.46
Beilstein:
1634065
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

<1 mmHg ( 20 °C)

Assay

98%

form

solid

bp

330 °C (lit.)

mp

43-45 °C (lit.)

functional group

amine

SMILES string

CCCCCCCCCCCCCCCCN

InChI

1S/C16H35N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17/h2-17H2,1H3

InChI key

FJLUATLTXUNBOT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Hexadecylamine (HDA) can be used:
  • To prepare a mixture of hexadecylamine-trioctylphosphine oxide-trioctylphosphine (HDA-TOPO-TOP) used in the synthesis of highly luminescent monodisperse CdSe and CdSe/ZnS nanocrystals.
  • As a capping agent in the synthesis of copper nanocrystals with different shapes by reducing CuCl2 with glucose.
  • As an organophilic alkylamine in organoclay in the preparation of a polyimide nanocomposite.
  • To form a membrane with vanadium oxide (V2O5/HDA) as the sensing layer of the extended gate H+-ion sensitive field effect transistor (EGFET) device.
  • As both reducing and stabilizing agent in the preparation of HDA capped Au and Ag nanoparticles applicable in surface-enhanced Raman scattering (SERS).
  • As a template in the synthesis of MCM-41 analog silica beads.
  • Along with lauric acid used in the preparation of cobalt (Co) nanorod superlattices with magnetic properties.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Asp. Tox. 1 - Eye Dam. 1 - Skin Corr. 1A - STOT RE 2

Target Organs

Gastrointestinal tract,Liver,Immune system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

285.8 °F - closed cup

Flash Point(C)

141 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Polyimide nanocomposite with a hexadecylamine clay: Synthesis and characterization.
Chang J-H, et al.
Journal of Applied Polymer Science, 84(12), 2294-2301 (2002)
1-Hexadecylamine as both reducing agent and stabilizer to synthesize Au and Ag nanoparticles and their SERS application
Hou X, et al.
Journal of Nanoparticle Research, 13(5), 1929-1936 (2011)
Synthesis and characterization of vanadium oxide/hexadecylamine membrane and its application as pH-EGFET sensor
Guerra EM and Mulato M
Journal of Sol-Gel Science and Technology, 52(3), 315-315 (2009)
Synthesis and magnetic properties of Co nanorod superlattices
Wetz F, et al.
Materials Science and Engineering, C, 27(5-8), 1162-1166 (2007)
Enyan Guo et al.
Physical chemistry chemical physics : PCCP, 17(1), 563-574 (2014-11-20)
We report on high-performance dye-sensitized solar cells (DSSCs) based on nitrogen doped anatase TiO2-CuxO core-shell mesoporous hybrids synthesized through a facile and controlled combined sol-gel and hydrothermal process in the presence of hexadecylamine as the structure-directing agent. The matching of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service