Skip to Content
Merck
All Photos(1)

Key Documents

648663

Sigma-Aldrich

Bis(ethylcyclopentadienyl)ruthenium(II)

Synonym(s):

Diethylruthenocene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C7H9RuC7H9
CAS Number:
Molecular Weight:
287.36
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

liquid

composition

Ru, 33.9-36.4% gravimetric

reaction suitability

core: ruthenium
reagent type: catalyst

refractive index

n20/D 1.5870 (lit.)

bp

100 °C/0.01 mmHg (lit.)

mp

6 °C (lit.)

density

1.3412 g/mL at 25 °C (lit.)

storage temp.

−20°C

SMILES string

[Ru].CC[C]1[CH][CH][CH][CH]1.CC[C]2[CH][CH][CH][CH]2

InChI

1S/2C7H9.Ru/c2*1-2-7-5-3-4-6-7;/h2*3-6H,2H2,1H3;

InChI key

VLTZUJBHIUUHIK-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Bis(ethylcyclopentadienyl)ruthenium(II) (Ru(EtCp)2), a metal organic is used as an atomic layer deposition precursor for Ru thin filmsand well-aligned RuO2 nanorods.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

>199.9 °F - closed cup

Flash Point(C)

> 93.3 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thermal atomic layer deposition (ALD) of Ru films for Cu direct plating.
Choi SH, et al.
Journal of the Electrochemical Society, 158(6), D351-D356 (2011)
In-situ real-time ellipsometric investigations during the atomic layer deposition of ruthenium: A process development from [(ethylcyclopentadienyl)(pyrrolyl) ruthenium] and molecular oxygen.
Knaut M, et al.
Journal of Vacuum Science & Technology. A, Vacuum, Surfaces, And Films, 30(1), 01A151-01A151 (2012)
Growth and characterization of the coexistence of vertically aligned and twinned V-shaped RuO2 nanorods on nanostructural TiO2 template
Chen CA, et al.
J. Alloy Compounds, 485(1-2), 524-528 (2009)
ALD-grown seed layers for electrochemical copper deposition integrated with different diffusion barrier systems.
Waechtler T, et al.
Microelectronic Engineering, 88(5), 684-689 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service