510475
Bis(methylcyclopentadienyl)nickel(II)
97%
Synonym(s):
1,1′-Dimethylnickelocene
About This Item
Recommended Products
Quality Level
Assay
97%
form
liquid
reaction suitability
core: nickel
reagent type: catalyst
bp
85-90 °C/1 mmHg (lit.)
mp
34-36 °C (lit.)
storage temp.
2-8°C
SMILES string
[Ni].C[C]1[CH][CH][CH][CH]1.C[C]2[CH][CH][CH][CH]2
InChI
1S/2C6H7.Ni/c2*1-6-4-2-3-5-6;/h2*2-5H,1H3;
InChI key
IHRNDXJDUYVDRB-UHFFFAOYSA-N
1 of 4
This Item | 700754 | SML3593 | T4069 |
---|---|---|---|
core: nickel, reagent type: catalyst | - | - | - |
liquid | solid | powder | solid |
2-8°C | −20°C | 2-8°C | room temp |
85-90 °C/1 mmHg (lit.) | - | - | - |
34-36 °C (lit.) | 198-203 °C | - | - |
Application
- Nickel sandwich complexes
- N-heterocyclic carbene complexes
- Functionally substituted monocyclopentadienyl compounds
Features and Benefits
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
150.8 °F - closed cup
Flash Point(C)
66 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.
Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.
Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.
Nickel complexes catalyze various synthetic reactions like oxidative addition, C-H activation, and cross-coupling.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service