Skip to Content
Merck
All Photos(1)

Key Documents

262455

Sigma-Aldrich

Bis(cyclopentadienyl)ruthenium(II)

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H10Ru
CAS Number:
Molecular Weight:
231.26
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

reaction suitability

core: ruthenium
reagent type: catalyst

mp

199-201 °C (lit.)

SMILES string

[Ru].[CH]1[CH][CH][CH][CH]1.[CH]2[CH][CH][CH][CH]2

InChI

1S/2C5H5.Ru/c2*1-2-4-5-3-1;/h2*1-5H;

InChI key

BKEJVRMLCVMJLG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

related product

Product No.
Description
Pricing

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Wenzel et al.
Strahlentherapie und Onkologie : Organ der Deutschen Rontgengesellschaft ... [et al], 162(1), 51-56 (1986-01-01)
The organ distribution of 103Ru labelled ruthenocenyl derivatives of tyramine, histamine, benzylamine, phenylethylamine and homoveratrylamine were measured in rats. The derivatives of tyramine, histamine and benzylamine showed a high affinity for the adrenal and ovar. Adrenal/muscle ratios up to 2000:1
Results of ERBF and ERPF measurements in healthy dogs with two new radiopharmaceutical principles.
R de Jong et al.
Contributions to nephrology, 56, 49-52 (1987-01-01)
Cynthia T Sanderson et al.
Inorganic chemistry, 44(9), 3283-3289 (2005-04-26)
Electronic absorption and resonance Raman spectral studies of benzoylruthenocene (BRc) and 1,1'-dibenzoylruthenocene (DRc) indicate that the low-energy electronic excited states of these 4d(6) metallocenes possess metal-to-ligand charge transfer (MLCT) character. While this MLCT contribution should weaken the metal-ring bonding in
Malay Patra et al.
Chemical communications (Cambridge, England), 47(41), 11444-11446 (2011-09-22)
A convenient synthesis of azidomethyl-ruthenocene and its use in the covalent labelling of amino acids, peptides and a peptide nucleic acid (PNA) monomer derivative by Cu(I) catalyzed azide-alkyne coupling (Cu-AAC, "click chemistry") are described.
M Wenzel et al.
International journal of radiation applications and instrumentation. Part A, Applied radiation and isotopes, 37(6), 491-495 (1986-01-01)
The potential radiopharmaceuticals: ruthenocenoyl alanine, ruthenocenoyl methionine, 1'-methyl-ruthenocenoyl glycine and its esters were labelled with 103Ru starting from the analogous ferrocene compounds. In a series of tests in mice and rats these substances were compared with hippuran and ruppuran (=

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service