Skip to Content
Merck
All Photos(2)

Key Documents

542032

Sigma-Aldrich

Diethyl(3-bromopropyl)phosphonate

95%

Synonym(s):

(3-Bromopropyl)phosphonic acid diethyl ester, NSC 138345

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)2POCH2CH2CH2Br
CAS Number:
Molecular Weight:
259.08
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

reaction suitability

reaction type: C-C Bond Formation

refractive index

n20/D 1.4620 (lit.)

bp

94-95 °C/2 mmHg (lit.)

density

1.348 g/mL at 25 °C (lit.)

SMILES string

CCOP(=O)(CCCBr)OCC

InChI

1S/C7H16BrO3P/c1-3-10-12(9,11-4-2)7-5-6-8/h3-7H2,1-2H3

InChI key

PEIKTSJIUKYDPC-UHFFFAOYSA-N

Application

Reactant for synthesis of:
  • Hybrid assembly composed of titanium dioxide nanoparticles and thin multi-walled carbon nanotubes
  • Organosoluble zirconium phosphonate nanocomposites for asymmetric hydrogenation
  • Cyclic hydroxamic acids via alkylation followed by intramolecular cyclization
  • pH-responsive cyclopolymer

Reactant for:
  • Homogenization of zirconium hydroxide phosphonate supported ruthenium catalyst
  • Reactions with sodium imidazolide

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service