Skip to Content
Merck
All Photos(1)

Key Documents

518751

Sigma-Aldrich

4-Aminophenylboronic acid pinacol ester

97%

Synonym(s):

2-(4-Aminophenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzeneamine, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)phenylamine, 4-Aminophenylboronic acid, pinacol cyclic ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H18BNO2
CAS Number:
Molecular Weight:
219.09
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

mp

165-169 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2ccc(N)cc2

InChI

1S/C12H18BNO2/c1-11(2)12(3,4)16-13(15-11)9-5-7-10(14)8-6-9/h5-8H,14H2,1-4H3

InChI key

ZANPJXNYBVVNSD-UHFFFAOYSA-N

Application

4-Aminophenylboronic acid pinacol ester can be used as a reagent for:
  • The preparation of substituted 3-phenyl-4H-1-benzopyran-4-ones by reacting with iodochromones via Pd catalyzed Suzuki-Miyaura cross-coupling reaction.
  • Mercury(II) detection by fluorometry with new fluorogenic indicators based on through-bond energy transfer from pentaquinone to rhodamine.
  • Rhodium-catalyzed amination reactions.
  • Palladium-catalyzed Suzuki cross-coupling to synthesize potential antitubercular and antimicrobial compounds.

It can also be used to prepare:
  • Hexaphenylbenzene derivatives as a potential bioprobe and multichannel keypad system.
  • Pyromellitic diimide-based polymer as matrix for solution-processable n-channel field-effect transistors.
  • Alternating copolymers of oligoarylenes and naphthalene bisimides as low band-gap semiconductors with electrochemical and spectroelectrochemical behavior.
  • γ-secretase modulators in the treatment of amyloid β formation.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis and Characterization of a Pyromellitic Diimide-Based Polymer with C- and N-Main Chain Links: Matrix for Solution-Processable n-Channel Field-Effect Transistors
Kola, S.; et al.
ACS Macro Letters, 1, 136-140 (2012)
Aminothiazoles as γ-secretase modulators
Luebbers, T.; et al.
Bioorganic & Medicinal Chemistry Letters, 21, 6554-6558 (2012)
Vandana Bhalla et al.
Organic letters, 14(4), 1012-1015 (2012-01-28)
Zinc ensemble of hexaphenylbenzene derivative 3 exhibits sensitive response toward adenosine monophosphate (AMP) and H(2)PO(4)(-) ions. Further, the application of derivative 3 as a multichannel molecular keypad could be realized in the presence of inputs of Zn(2+) ions, H(2)PO(4)(-) ions
David A Vasselin et al.
Journal of medicinal chemistry, 49(13), 3973-3981 (2006-06-23)
A new series of fluoro-, methoxyl-, and amino-substituted isoflavones have been synthesized as potential antitumor agents based on structural similarities to known flavones and isoflavones (quercetin and genistein respectively) and antitumor 2-phenylbenzothiazoles. Target compounds were synthesized using palladium-catalyzed coupling methodologies
Michael König et al.
Tetrahedron, 67(23), 4243-4252 (2011-07-16)
The palladium-catalyzed Suzuki-Miyaura cross-coupling reaction has been investigated on meso-substituted trans-A(2)B-corrole using tailored Pd-catalyst systems.We present the first examples of Suzuki-Miyaura cross-coupling reactions on meso-substituted trans-A(2)B-corrole derivatives with neutral, sterically hindered, inactivated and heteroaromatic boronic acids and esters, alkenylboronic acids

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service