Skip to Content
Merck
All Photos(2)

Key Documents

523968

Sigma-Aldrich

3-Hydroxyphenylboronic acid

≥95.0%

Synonym(s):

3-Hydroxybenzeneboronic acid, m-Hydroxybenzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4B(OH)2
CAS Number:
Molecular Weight:
137.93
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95.0%

mp

210-213 °C (dec.) (lit.)

SMILES string

OB(O)c1cccc(O)c1

InChI

1S/C6H7BO3/c8-6-3-1-2-5(4-6)7(9)10/h1-4,8-10H

InChI key

WFWQWTPAPNEOFE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-Hydroxyphenylboronic acid (3-HPBA) can be used as a reagent:
  • In Suzuki-Miyaura coupling reactions with aryl halides for the formation of C-C bond in the presence of Pd catalyst.[1][2]
  •  To synthesize boron/nitrogen-doped polymer nano/microspheres by hydrothermal polymerization with formaldehyde and ammonia.[3]      
  • To prepare carbon quantum dots based on 3-HPBA as selective fructose sensor.[4]     
  • In the development of modified electrodes for electrochemical biosensors.[5]

Footnote

Contains varying amounts of anhydride

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana M A Crista et al.
Journal of fluorescence, 29(1), 265-270 (2019-01-07)
The selective fluorescence sensing of fructose was achieved by fluorescence quenching of the emission of hydrothermal-synthesized carbon quantum dots prepared by 3-hydroxyphenylboronic acid. Quantification of fructose was possible in aqueous solutions with pH of 9 (Limit of Detection LOD and
Facile synthesis of monodisperse bulk boron-and nitrogen-doped carbon nano/microspheres
Zhao J, et al.
Journal of Material Chemistry A, 6(46), 23780-23786 (2018)
Synthesis of hyperbranched polythiophene with a controlled degree of branching via catalyst-transfer Suzuki-Miyaura coupling reaction
Segawa Y, et al.
Polym. Chem., 4(4), 1208-1215 (2013)
Mingyan Zhu et al.
ACS combinatorial science, 14(2), 124-134 (2011-12-21)
As a continuation of our previous report (J. Comb. Chem.2010, 12, 548-558), we accomplished the diversity-oriented synthesis of polyheterocyclic small-molecule library with privileged benzopyran substructure. To ensure the synthetic efficiency, we utilized the solid-phase parallel platform and the fluorous-tag-based solution-phase
Recent progress in electrochemical biosensors based on phenylboronic acid and derivatives
Anzai J-i
Materials Science and Engineering, C, 67, 737-746 (2016)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service