Skip to Content
Merck
All Photos(2)

Key Documents

441651

Sigma-Aldrich

3-Formylphenylboronic acid

≥95%

Synonym(s):

(3-Formylbenzene)boronic acid, 3-(Dihydroxyboryl)benzaldehyde, 3-Boronobenzaldehyde, m-Formylphenylboronic acid, m-formyl-benzeneboronic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HCOC6H4B(OH)2
CAS Number:
Molecular Weight:
149.94
Beilstein:
3030769
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

≥95%

form

powder

mp

109-113 °C

functional group

aldehyde

SMILES string

OB(O)c1cccc(C=O)c1

InChI

1S/C7H7BO3/c9-5-6-2-1-3-7(4-6)8(10)11/h1-5,10-11H

InChI key

HJBGZJMKTOMQRR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reagent used:
  • To study the effects of boronic acid on fluoride-selective chemosignaling behavior of merocyanine dye
  • As exciton-coupled CD probes for epigallocatechin gallate

Biological inhibitor of γ-glutamyltranspeptidase

Reactant involved in:
  • Palladium-catalyzed homocoupling
  • Suzuki coupling reactions

Other Notes

Contains varying amounts of anhydride

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mijun Peng et al.
Journal of chromatography. A, 1474, 8-13 (2016-11-09)
Rapid and efficient extraction of bioactive glycosides from complex natural origins poses a difficult challenge, and then is often inherent bottleneck for their highly utilization. Herein, we propose a strategy to fabricate boronate affinity based surface molecularly imprinted polymers (MIPs)
Hua-Wei Liu et al.
Chemistry, an Asian journal, 12(13), 1545-1556 (2017-04-19)
Sialic acids play important roles in mammalian development, cell-cell attachment, and signaling. As cancer cells utilize their overexpressed sialylated antigens to propagate metastases, the development of probes for sialic acids is of high importance. Herein, we report three luminescent cyclometalated

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service