Skip to Content
Merck
All Photos(1)

Key Documents

490040

Sigma-Aldrich

Sodium L-lactate-3-13C solution

45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral Purity, HPLC)

Synonym(s):

L-Lactic acid-3-13C sodium salt solution

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
13CH3CH(OH)CO2Na
CAS Number:
Molecular Weight:
113.05
MDL number:
UNSPSC Code:
12352106
PubChem Substance ID:
NACRES:
NA.12

description

Packaged as gram of solution, 45-55% (w/w)

Quality Level

isotopic purity

≥99 atom % 13C

Assay

≥98% (CP)
≥98% (Chiral Purity, HPLC)

concentration

45-55 % (w/w) in H2O

technique(s)

mass spectrometry (MS): suitable

refractive index

n20/D 1.42

mass shift

M+1

storage temp.

−20°C

SMILES string

[Na+].[13CH3][C@H](O)C([O-])=O

InChI

1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i1+1;

InChI key

NGSFWBMYFKHRBD-UEORGECYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Sodium L-lactate-3-13C is an isotopic analogue of Sodium L-lactate.

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Glöggler et al.
Scientific reports, 6, 36080-36080 (2016-11-05)
In order to study metabolic processes in animal models of diseases and in patients, microdialysis probes have evolved as powerful tools that are minimally invasive. However, analyses of microdialysate, performed remotely, do not provide real-time monitoring of microdialysate composition. Microdialysate
Robin Wijngaard et al.
International journal of molecular sciences, 22(9) (2021-05-06)
The metabolic ratios lactate/pyruvate and β-hydroxybutyrate/acetoacetate are considered valuable tools to evaluate the in vivo redox cellular state by estimating the free NAD+/NADH in cytoplasm and mitochondria, respectively. The aim of the current study was to validate a gas-chromatography mass
Hua Qing et al.
Cell, 182(2), 372-387 (2020-07-02)
Acute psychological stress has long been known to decrease host fitness to inflammation in a wide variety of diseases, but how this occurs is incompletely understood. Using mouse models, we show that interleukin-6 (IL-6) is the dominant cytokine inducible upon

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service