486191
Sodium pyruvate-2,3-13C2
99 atom % 13C
Synonym(s):
α-Ketopropionic acid-2,3-13C2 sodium salt, 2-Oxopropanoic acid-2,3-13C2 sodium salt, 13C Labeled sodium pyruvate, Pyruvic acid-2,3-13C2 sodium salt
About This Item
Recommended Products
isotopic purity
99 atom % 13C
Quality Level
form
solid
technique(s)
mass spectrometry (MS): suitable
mp
>300 °C (lit.) (lit.)
mass shift
M+2
storage temp.
2-8°C
SMILES string
[Na+].[13CH3][13C](=O)C([O-])=O
InChI
1S/C3H4O3.Na/c1-2(4)3(5)6;/h1H3,(H,5,6);/q;+1/p-1/i1+1,2+1;
InChI key
DAEPDZWVDSPTHF-AWQJXPNKSA-M
Related Categories
Packaging
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Sens. 1B
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Articles
Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.
Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.
Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.
Review the 10 steps of glycolysis in the Embden-Meyerhof-Parnas glycolytic pathway. Easily compare reaction stages and buy the enzymes for your life science research.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service