589209
Sodium L-lactate-2-13C solution
45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral Purity, HPLC)
Synonym(s):
L-Lactic acid-2-13C sodium salt solution
About This Item
Recommended Products
description
Packaged as gram of solution, 45-55% (w/w) solution
isotopic purity
≥99 atom % 13C
Assay
≥98% (CP)
≥98% (Chiral Purity, HPLC)
concentration
45-55 % (w/w) in H2O
technique(s)
mass spectrometry (MS): suitable
mp
163-165 °C (lit.)
mass shift
M+1
storage temp.
−20°C
SMILES string
[Na+].C[13C@H](O)C([O-])=O
InChI
1S/C3H6O3.Na/c1-2(4)3(5)6;/h2,4H,1H3,(H,5,6);/q;+1/p-1/t2-;/m0./s1/i2+1;
InChI key
NGSFWBMYFKHRBD-LHMCEBHZSA-M
Looking for similar products? Visit Product Comparison Guide
Related Categories
Packaging
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service