Merck
Origin and Function of Stress-Induced IL-6 in Murine Models.
Cell (2020-07-02)
Hua Qing, Reina Desrouleaux, Kavita Israni-Winger, Yann S Mineur, Nia Fogelman, Cuiling Zhang, Saleh Rashed, Noah W Palm, Rajita Sinha, Marina R Picciotto, Rachel J Perry, Andrew Wang
ABSTRACT
Acute psychological stress has long been known to decrease host fitness to inflammation in a wide variety of diseases, but how this occurs is incompletely understood. Using mouse models, we show that interleukin-6 (IL-6) is the dominant cytokine inducible upon acute stress alone. Stress-inducible IL-6 is produced from brown adipocytes in a beta-3-adrenergic-receptor-dependent fashion. During stress, endocrine IL-6 is the required instructive signal for mediating hyperglycemia through hepatic gluconeogenesis, which is necessary for anticipating and fueling "fight or flight" responses. This adaptation comes at the cost of enhancing mortality to a subsequent inflammatory challenge. These findings provide a mechanistic understanding of the ontogeny and adaptive purpose of IL-6 as a bona fide stress hormone coordinating systemic immunometabolic reprogramming. This brain-brown fat-liver axis might provide new insights into brown adipose tissue as a stress-responsive endocrine organ and mechanistic insight into targeting this axis in the treatment of inflammatory and neuropsychiatric diseases.
Sigma-Aldrich
CL 316,243 hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Sodium L-lactate-3-13C solution, 45-55 % (w/w) in H2O, ≥99 atom % 13C, ≥98% (CP), ≥98% (Chiral Purity, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
Poloxamer 407, purified, non-ionic
Sigma-Aldrich
6-Hydroxydopamine hydrochloride, ≥97% (titration), powder
Sigma-Aldrich
(±)-Propranolol hydrochloride, ≥99% (TLC), powder
USP
Isoproterenol hydrochloride, United States Pharmacopeia (USP) Reference Standard