Skip to Content
Merck
All Photos(3)

Key Documents

463744

Sigma-Aldrich

Vanadium(III) oxide

greener alternative

99.99% trace metals basis

Synonym(s):

Divanadium trioxide, Vanadic oxide, Vanadium sesquioxide, Vanadium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
V2O3
CAS Number:
Molecular Weight:
149.88
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.99% trace metals basis

form

powder and chunks

composition

V2O3

reaction suitability

reagent type: catalyst
core: vanadium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

mp

1970 °C (lit.)

density

4.87 g/mL at 25 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

O=[V]O[V]=O

InChI

1S/3O.2V

InChI key

KFAFTZQGYMGWLU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Click here for more details.

Application

Vanadium(III) oxide (V2O3) can be used for a variety of sustainable applications such as:
  • anode material for the fabrication of lithium-ion batteries
  • fabrication of vanadium redox flow batteries
  • preparation of vanadium nitride (VN) for potential usage in hydrodenitrogenation (HDN), and hydrodeoxygenation (HDO)

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Electrochemical performance investigation of electrospun urchin-like V2O3-CNF composite nanostructure for vanadium redox flow battery
Busacca C, et al.
Electrochimica Acta, 230(1), 174-180 (2017)
Electrochemical characteristics of lithium vanadate, Li1+ xVO2, new anode materials for lithium ion batteries
Song JH, et al.
Journal of Power Sources, 195(18), 6157-6161 (2010)
Preparation of Vanadium Nitride by Magnesiothermic Reduction of V 2 O 3 in Nitrogen Atmosphere
Wu Y, et al.
Metall. Mater. Trans. B, 49(6), 3570-3579 (2018)

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service