Skip to Content
Merck
All Photos(3)

Key Documents

215066

Sigma-Aldrich

Gallium(III) oxide

≥99.99% trace metals basis

Synonym(s):

Gallium trioxide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Ga2O3
CAS Number:
Molecular Weight:
187.44
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

≥99.99% trace metals basis

form

(crystalline powder)

reaction suitability

reagent type: catalyst
core: gallium

density

5.88 g/mL at 25 °C

SMILES string

O=[Ga]O[Ga]=O

InChI

1S/2Ga.3O

InChI key

QZQVBEXLDFYHSR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gallium(III) oxide (Ga2O3) is a wide band gap semiconductor that belongs to a family of transparent semiconducting oxides (TSO). It can form different polymorphs such as α-,β-, γ-, δ-, and ε-. Polycrystalline and nanocrystalline Ga2O3 can be prepared using several methods such as chemical vapor deposition, thermal vaporization, and sublimation, molecular beam epitaxy, melt growth, etc. It is widely used as a functional material in various applications including optoelectronics, chemical sensors, catalysis, semiconductor devices, field-effect transistors, and many others.

Application

Ga2O3 is widely used as a host material for the fabrication of electroluminescent devices. For example, europium-doped Ga2O3 thin films can be used as a light-emitting layer to fabricate an optically transparent electroluminescent device.

Due to its distinct optical and electrical properties like moderate conductivity and high laser damage threshold, Ga2O3 can be used in laser-driven electron accelerators, low-loss plasmonics, and Si-based dielectric laser accelerators.

It can also be used as an effective catalyst for the dehydrogenation of propane to propene.
Starting material for the preparation of Sr2CuGaO3S, an example of a rare square pyramidal gallium.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Naoya Kumagai et al.
Chemical communications (Cambridge, England), 47(6), 1884-1886 (2010-12-07)
Using the Rh(3+) ion (Rh d(6)) in a regular octahedral coordination, which forms fully occupied t(2g)(6) and empty e(g)(0) as a result of ligand-field splitting, we demonstrated that Rh-doped ZnGa(2)O(4) had midgap states created by t(2g)(6) and e(g)(0) that had
Bipin Pandey et al.
Langmuir : the ACS journal of surfaces and colloids, 28(38), 13705-13711 (2012-09-01)
This paper reports the formation of self-organized nanoporous gallium oxide by anodization of solid gallium metal. Because of its low melting point (ca. 30 °C), metallic gallium can be shaped into flexible structures, permitting the fabrication of nanoporous anodic oxide
Baoxiu Zhao et al.
Journal of environmental sciences (China), 24(4), 774-780 (2012-08-17)
Perfluorooctanoic acid (PFOA) is a new-found hazardous persistent organic pollutant, and it is resistant to decomposition by hydroxyl radical (HO*) due to its stable chemical structure and the high electronegativity of fluorine. Photocatalytic reduction of PFOA with beta-Ga2O3 in anoxic
Vladimir N Sigaev et al.
Nanoscale, 5(1), 299-306 (2012-11-21)
Nanoparticles in amorphous oxides are a powerful tool for embedding a wide range of functions in optical glasses, which are still the best solutions in several applications in the ever growing field of photonics. However, the control of the nanoparticle
The rate of charge tunneling through self-assembled monolayers is insensitive to many functional group substitutions.
Hyo Jae Yoon et al.
Angewandte Chemie (International ed. in English), 51(19), 4658-4661 (2012-04-17)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service