Skip to Content
Merck
All Photos(3)

Key Documents

415138

Sigma-Aldrich

Strontium oxide

greener alternative

99.9% trace metals basis

Synonym(s):

Strontia, Strontium monoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SrO
CAS Number:
Molecular Weight:
103.62
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

powder

reaction suitability

reagent type: catalyst
core: strontium

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

density

4.7 g/mL at 25 °C (lit.)

greener alternative category

SMILES string

O=[Sr]

InChI

1S/O.Sr

InChI key

UFQXGXDIJMBKTC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency and is intended for Molecular Solar Thermal Energy Storage Systems (MOST). Click here for more information.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Corr. 1B

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ajay Kumar Choudhary et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 185, 155-162 (2017-06-02)
Er
Barış Demirel et al.
Materials (Basel, Switzerland), 13(21) (2020-11-01)
This study was aimed to produce and characterize the first commercial glass materials with enhanced antibacterial property using conventional melting method. For this purpose, typical container glass composition that contains some specific metal ions, such as silver, strontium, and copper
Biao Hu et al.
Tissue & cell, 66, 101386-101386 (2020-09-17)
Ideal bone defect repair scaffolds should be biodegradable, biocompatible, bioactive, porous, and provide adequate mechanical support. However, it is challenging to fabricate such an ideal bone repair scaffold. Previously, we showed that 5 wt.% strontium-doped hydroxyapatite (Sr-HA) scaffolds prepared by
Yung-Cheng Chiu et al.
International journal of molecular sciences, 20(11) (2019-06-06)
In this study, we synthesized strontium-contained calcium silicate (SrCS) powder and fabricated SrCS scaffolds with controlled precise structures using 3D printing techniques. SrCS scaffolds were shown to possess increased mechanical properties as compared to calcium silicate (CS) scaffolds. Our results
Jean-Marc Tulliani et al.
Sensors (Basel, Switzerland), 13(9), 12070-12092 (2013-09-13)
The aim of this work is to study the sensing behavior of Sr-doped hematite for soil water content measurement. The material was prepared by solid state reaction from commercial hematite and strontium carbonate heat treated at 900 °C. X-Ray diffraction

Articles

As with all types of fuel cells, a Solid Oxide Fuel Cell (SOFC) is capable of efficiently transforming chemical energy into electrical energy.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

Three approaches generate white light, including LED-based down-conversion for broader applications.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service