Skip to Content
Merck
All Photos(1)

Key Documents

409308

Sigma-Aldrich

Tin(II) iodide

AnhydroBeads, −10 mesh, 99.99% trace metals basis

Synonym(s):

Stannous iodide, Tin diiodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
SnI2
CAS Number:
Molecular Weight:
372.52
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.99% trace metals basis

form

beads

impurities

≤150.0 ppm Trace Metal Analysis

particle size

−10 mesh

bp

714 °C (lit.)

mp

320 °C (lit.)

density

5.28 g/mL at 25 °C (lit.)

SMILES string

I[SnH2]I

InChI

1S/2HI.Sn/h2*1H;/q;;+2/p-2

InChI key

JTDNNCYXCFHBGG-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Tin(II) iodide AnhydroBeads−10 mesh, 99.99% trace metals basis comes as beads with red to purple in color with applications in semiconductor research, solar cells, material science, chemical synthesis, catalysis, and photonics. Tin (II) iodide is widely used as a precursor toprepare lead-free non-toxic hybrid perovskite materials. Tin-based perovskitesshow excellent electrical and optical properties such as high charge carrier mobility,absorption coefficient, and small exciton binding energies.

Application

Tin(II) iodide (SnI₂) is a versatile compound with a range of applications in research, particularly in semiconductor technology, solar cells, chemical synthesis, catalysis etc. SnI₂ is used in perovskite solar cells as a precursor for tin-based perovskites or as an additiveto improve device stability and performance. The addition of a small amount of 2D tin film induces well-defined orientation and superior crystallinity in formamidinium tin iodide (FASnI3) films. This results in the longer life of charge carriers and improves the performance of hybrid perovskite solar cell (HPSC). [Ref] It can also be used to preparesolution-processable lamellar hybrid [CH3(CH2)11NH3]SnI3semiconductor.Its catalytic properties can be leveraged to develop new synthetic methodologies such as reductions, cyclizations, and coupling reactions. It is suitable to be used in photonic applications, including sensors and photovoltaic devices. It is used as a deposition material for preparing thin films for use in electronic and optoelectronic devices. Techniques like chemical vapor deposition (CVD) and physical vapor deposition (PVD) are explored for creating high-quality films. In a study, it is found that when a novel catalytic system comprised of tin sulfide (SnS) nanoflakes as a solid catalyst and tin iodide (SnI2) as a dual-functional electrolyte additive, the Li-air battery enables operating at high current rates up to 10 000 mA g−1 (corresponding to 1 mA cm−2). Also it has been observed that that the role of the SnI2 is not only reacting with the lithium anode to provide protection but reducing the charge potential by promoting catalytic decomposition of the Li2O2.

Features and Benefits

Tin(II) iodide AnhydroBeads, −10 mesh, 99.99% trace metals basis is designed and tested under stringent dry manufacturing conditions to ensure low water content, trace metal purity of 99.99%, and low surface area-to-volume ratio. The salt possesses excellent electrical and optical properties such as high charge carrier mobility,absorption coefficient, and small exciton binding energies. The advantages of our AnhydroBeads salts are as follows:

1) Reduced uptake rate of environmental moisture minimizes caking, dusting, and static buildup for repeated easy handling.

2) Higher crucible packing densities and lower volatility in high-temperature solid state procedures.

3) Easier pneumatic loading of salts to sample chambers due to less clogging issues associated with powdered salt counterparts.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Dam. 1 - Met. Corr. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Cardio-vascular system,hematopoietic system, Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Perovskite multifunctional logic gates via bipolar photoresponse of single photodetector
Kim W, et al.
Nature Communications, 13, 720-720 (2022)
Novel Co-Catalytic Activities of Solid and Liquid Phase Catalysts in High-Rate Li-Air Batteries
Zhang C, et al.
Advanced Energy Materials, 12, 2201616-2201616 (2022)
Liang, K. et al.
Chemistry of Materials, 10, 403-403 (1998)
A Cation-Exchange Approach for the Fabrication of Efficient Methylammonium Tin Iodide Perovskite Solar Cells
Angewandte Chemie (International ed. in English), 58, 6688-6692 (2019)
Kohei Nishimura et al.
ACS applied materials & interfaces, 11(34), 31105-31110 (2019-08-07)
In the composition of Q0.1(FA0.75MA0.25)0.9SnI3, Q is replaced with Na+, K+, Cs+, ethylammonium+ (EA+), and butylammonium+ (BA+), respectively, and the relationship between actually measured lattice strain and photovoltaic performances is discussed. The lattice strain evaluated by the Williamson-hall plot of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service