Merck
  • Relationship between Lattice Strain and Efficiency for Sn-Perovskite Solar Cells.
Relationship between Lattice Strain and Efficiency for Sn-Perovskite Solar Cells.
ACS applied materials & interfaces (2019-08-07)
Kohei Nishimura, Daisuke Hirotani, Muhammad Akmal Kamarudin, Qing Shen, Taro Toyoda, Satoshi Iikubo, Takashi Minemoto, Kenji Yoshino, Shuzi Hayase
ABSTRACT
In the composition of Q0.1(FA0.75MA0.25)0.9SnI3, Q is replaced with Na+, K+, Cs+, ethylammonium+ (EA+), and butylammonium+ (BA+), respectively, and the relationship between actually measured lattice strain and photovoltaic performances is discussed. The lattice strain evaluated by the Williamson-hall plot of X-ray diffraction data decreased as the tolerance factor was close to one. The efficiency of the Sn-perovskite solar cell was enhanced as the lattice strain decreased. Among them, EA0.1(FA0.75MA0.25)0.9SnI3 having lowest lattice strain gave the best result of 5.41%. Because the carrier mobility increased with a decrease in the lattice strain, these lattice strains would disturb carrier mobility and decrease the solar cell efficiency. Finally, the results that the efficiency of the SnGe-perovskite solar cells was gradually enhanced from 6.42 to 7.60% during storage, was explained by the lattice strain relaxation during the storage.
Sigma-Aldrich
Germanium(II) iodide, ≥99.8% trace metals basis
Sigma-Aldrich
Formamidinium iodide
Sigma-Aldrich
Methylammonium iodide, 98%
Sigma-Aldrich
n-Butylammonium iodide
Sigma-Aldrich
Ethylammonium Iodide
Sigma-Aldrich
Tin(II) fluoride, 99%