Skip to Content
Merck
All Photos(2)

Key Documents

203017

Sigma-Aldrich

Cesium bromide

99.999% trace metals basis

Synonym(s):

Cesium monobromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CsBr
CAS Number:
Molecular Weight:
212.81
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.999% trace metals basis
form:
powder and chunks
Pricing and availability is not currently available.

Quality Level

Assay

99.999% trace metals basis

form

powder and chunks

impurities

≤15.0 ppm Trace Metal Analysis

mp

636 °C (lit.)

density

4.44 g/mL at 25 °C (lit.)

SMILES string

[Br-].[Cs+]

InChI

1S/BrH.Cs/h1H;/q;+1/p-1

InChI key

LYQFWZFBNBDLEO-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Cesium bromide (CsBr) is an inorganic compound moderately soluble in water and exhibits hygroscopic properties. CsBr crystals are utilized in scintillation counters for detecting radiation due to their effective scintillation properties. It is also used in optics as a beam splitter component in wide-band spectrophotometers.

Application

Cesium bromide can be used as:      
  • An electrolyte additives to improve the cycling efficiency of lithium and graphite anodes in lithium-ion batteries. The cesium cations can help control the interfacial chemistry on the anode, leading to enhanced performance.     
  • An additive to enhance the efficiency and stability of CsPbI2Br perovskite solar cells by passivating defects, improving charge transport properties, and contributing to higher power conversion efficiencies.
  • As a precursor material to synthesize cesium lead bromide (CsPbBr3) perovskite, which is then utilized as an electrode material in a symmetric supercapacitor.  
  • As a precursor to prepare thermal neutron imaging plates.     
  • As a precursor to synthesize lead-free CsBr: Eu2+ nanocrystals for fabricating white LEDs.    
  • As an additive to prepare CsI-based scintillators. Alloying with cesium bromide increases the yield limit and microhardness of CsI crystals.

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Repr. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Caesium-binding sites in the gramicidin pore.
D A Doyle et al.
Biochemical Society transactions, 22(4), 1043-1045 (1994-11-01)
N Kurata et al.
Radiation protection dosimetry, 119(1-4), 398-401 (2006-06-01)
CsBr phosphor ceramics doped with different luminescence centres such as In2O3, Eu2O3, EuCl3, SmCl3, TbCl3, GdCl3 or NdCl3 as the candidate for a new optically-stimulable phosphor for medical X-ray imaging sensor were prepared using a conventional ceramic fabrication process. It
Peng Pan et al.
Analytical chemistry, 75(20), 5468-5474 (2004-01-09)
Solutions consisting of protein and small molecule mixtures have been subjected to electrospray ionization to study the influence of small molecule/cation components at high concentrations on the electrospray responses of proteins. Emphasis was placed on solutions consisting of equal parts
Matthieu P Wintergerst et al.
Journal of the American Chemical Society, 130(12), 4129-4139 (2008-03-04)
Solvent-extraction studies provide confirming evidence that meso-octamethylcalix[4]pyrrole acts as an ion-pair receptor for cesium chloride and cesium bromide in nitrobenzene solution. The stoichiometry of the interaction under extraction conditions from water to nitrobenzene was determined from plots of the cesium
Wen-Chen Zheng et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 64(3), 628-630 (2006-01-03)
The complete high-order perturbation formula of g factor, including not only the widely used crystal-field (CF) mechanism, but also the neglected change-transfer (CT) mechanism in the CF theory, is established for a 3d8 ion in cubic octahedral site. From the

Articles

Colloidal quantum dots (CQDs) are semiconducting crystals of only a few nanometers (ca. 2–12 nm) coated with ligand/surfactant molecules to help prevent agglomeration.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service