Skip to Content
Merck
All Photos(1)

Key Documents

329711

Sigma-Aldrich

Zinc dimethyldithiocarbamate

97%

Synonym(s):

Dimethyldithiocarbamic acid zinc salt, Ziram

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2NCSSZnSCSN(CH3)2
CAS Number:
Molecular Weight:
305.82
Beilstein:
3707008
EC Number:
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

reaction suitability

core: zinc
reagent type: catalyst

mp

248-257 °C (lit.)

SMILES string

CN(C)C(=S)S[Zn]SC(=S)N(C)C

InChI

1S/2C3H7NS2.Zn/c2*1-4(2)3(5)6;/h2*1-2H3,(H,5,6);/q;;+2/p-2

InChI key

DUBNHZYBDBBJHD-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Classifications

Acute Tox. 2 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1 - Skin Sens. 1 - STOT RE 2 Oral - STOT SE 3

Target Organs

Liver,Blood, Respiratory system

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Josiane P Lafleur et al.
Lab on a chip, 12(22), 4651-4656 (2012-07-25)
Conventional methods of environmental analysis can be significantly improved by the development of portable microscale technologies for direct in-field sensing at remote locations. This report demonstrates the vast potential of gold nanoparticle-based microfluidic sensors for the rapid, in-field, detection of
Anthony Wang et al.
European journal of epidemiology, 26(7), 547-555 (2011-04-21)
Due to the heavy and expanding agricultural use of neurotoxic pesticides suspected to affect dopaminergic neurons, it is imperative to closely examine the role of pesticides in the development of Parkinson's disease (PD). We focus our investigation on pesticide use
Gha-Young Kim et al.
Journal of environmental monitoring : JEM, 10(5), 632-637 (2008-05-02)
A highly sensitive enzyme electrode was prepared based on gold nanoparticles for measurement of pesticides. Gold nanoparticles of 25-30 nm were synthesized on a glassy carbon electrode by double-pulse technique while the coverage was controlled by applied potential and time.
Emanuela Corsini et al.
Toxicology and applied pharmacology, 212(2), 89-98 (2005-08-23)
Mancozeb, a polymeric complex of manganese ethylenebisdithiocarbamate with zinc salt, is widely used in agriculture as fungicide. Literature data indicate that ethylenebisdithiocarbamates (EBDTCs) may have immunomodulatory effects in humans. We have recently found in agricultural workers occupationally exposed to the
Arthur P Chou et al.
The Journal of biological chemistry, 283(50), 34696-34703 (2008-09-27)
The etiology of Parkinson disease (PD) is unclear but may involve environmental toxins such as pesticides leading to dysfunction of the ubiquitin proteasome system (UPS). Here, we measured the relative toxicity of ziram (a UPS inhibitor) and analogs to dopaminergic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service