Skip to Content
Merck
All Photos(1)

Key Documents

45707

Supelco

Zineb

PESTANAL®, analytical standard

Synonym(s):

Zinc-ethylenebis(dithiocarbamate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H6N2S4Zn
CAS Number:
Molecular Weight:
275.75
Beilstein:
4165797
EC Number:
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

analytical standard

Quality Level

description

technical mixture

product line

PESTANAL®

shelf life

limited shelf life, expiry date on the label

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

agriculture
environmental

format

neat

storage temp.

2-8°C

SMILES string

S=C1NCCNC(=S)S[Zn]S1

InChI

1S/C4H8N2S4.Zn/c7-3(8)5-1-2-6-4(9)10;/h1-2H2,(H2,5,7,8)(H2,6,9,10);/q;+2/p-2

InChI key

AMHNZOICSMBGDH-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Zineb belongs to the ethylenebisdithiocarbamates (EBDCs) class of compounds which has been employed in crop protection.

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
Zineb may be used as the reference standard for the determination of zineb in wheat samples by capillary electrophoresis with ultraviolet (UV) detection, in commercial formulations by flow injection analysis-atomic absorption spectrometry (FIA-AAS), environmental water samples by liquid chromatography-mass spectrometry (LC-MS) in scanning or selected ion monitoring (SIM) mode and in synthetic samples, spiked water samples and crops by H-point standard addition method (HPSAM) combined with ultraviolet-visible (UV-Vis) spectrophotometry.

Recommended products

Find a digital Reference Material for this product available on our online platform ChemisTwin® for NMR. You can use this digital equivalent on ChemisTwin® for your sample identity confirmation and compound quantification (with digital external standard). An NMR spectrum of this substance can be viewed and an online comparison against your sample can be performed with a few mouseclicks. Learn more here and start your free trial.

Legal Information

PESTANAL is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dinis Reis Oliveira et al.
Biofouling, 36(3), 332-350 (2020-05-14)
Today, ship hull fouling is managed through fouling-control coatings, complemented with in-water cleaning. During cleaning, coating damage and wear must be avoided, for maximum coating lifetime and reduced antifoulant release. When possible, cleaning should target early stages of fouling, using
LC/MS studies on characterization and determination of N,N′-ethylenebisdithiocarbamate fungicides in environmental water samples.
Hanada Y, et al.
Analytical Sciences, 18(4), 441-444 (2002)
H-Point standard addition method for simultaneous determination of maneb and zineb.
Kaur PP and Gupta U
Journal of Chemistry, 6(1), 106-112 (2009)
Capillary electrophoretic determination of zinc dimethyldithiocarbamate (Ziram) and zinc ethylenebisdithiocarbamate (Zineb).
Malik AK and Faubel W
Talanta, 52(2), 341-346 (2000)
Flow injection/atomic absorption spectrometric determination of zineb in commercial formulations of pesticide based on slurry sampling.
Cassella RJ, et al.
Analytical Sciences, 18(11), 1253-1256 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service