325902
Scandium(III) nitrate hydrate
99.9% trace metals basis
Synonym(s):
Scandium trinitrate hydrate, Scandium(3+) trinitrate hydrate, Trinitric acid scandium salt hydrate
About This Item
Recommended Products
Quality Level
Assay
99.9% trace metals basis
form
powder and chunks
composition
Degree of hydration, ~4
impurities
≤1500.0 ppm Trace Rare Earth Analysis
SMILES string
CO[N+]([O-])=O.CO[N+]([O-])=O.CO[N+]([O-])=O.[H]O[H].[Sc]
InChI
1S/3CH3NO3.H2O.Sc/c3*1-5-2(3)4;;/h3*1H3;1H2;
InChI key
QXEUKQIDJOLTAC-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Application
- For the fabrication of dense and crack-free scandium-stabilized zirconia (ScSZ) thin films by electrostatic spray deposition technique.
- To prepare scandium-incorporated indium oxide semiconductor which acts as an electron transport layer in a light-emitting field-effect transistor (LEFET).
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Ox. Sol. 2
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service