Skip to Content
Merck
All Photos(3)

Key Documents

229415

Sigma-Aldrich

Aluminum nitrate nonahydrate

99.997% trace metals basis

Synonym(s):

Aluminium nitrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Al(NO3)3 · 9H2O
CAS Number:
Molecular Weight:
375.13
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.997% trace metals basis

form

crystals and lumps

impurities

≤35.0 ppm Trace Metal Analysis

bp

135 °C (lit.)

mp

73 °C (lit.)

solubility

ethanol: very soluble(lit.)

application(s)

battery manufacturing

SMILES string

O.O.O.O.O.O.O.O.O.[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/Al.3NO3.9H2O/c;3*2-1(3)4;;;;;;;;;/h;;;;9*1H2/q+3;3*-1;;;;;;;;;

InChI key

SWCIQHXIXUMHKA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Aluminum nitrate nonahydrate is a crystalline aluminum salt of nitric acid primarily used as a catalyst in manufacturing petroleum products, including gasoline and other fuels. It is also used in the production of aluminum oxide and the fabrication of cathode material for lithium-ion batteries.

Application

Aluminum nitrate nonahydrate can be used as a precursor to synthesize:
  • AlF3-coated LiCoO2, which is used to prepare a blend composite cathode with Li [Ni1/3Co1/3Mn1/3] O2 for Li-ion batteries.
  • Disperse alumina.
  • Alumina gel by sol-gel method and pure gamma alumina by hydrolysis.
  • Nanocrystalline ZnAl2O4 by hydrothermal method.
  • Magnesium aluminum layered hydroxides which are applicable as nanocontainers of corrosion inhibitors for protective organic coatings.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

does not flash

Flash Point(C)

does not flash

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stefan Neatu et al.
Materials (Basel, Switzerland), 12(11) (2019-06-05)
This study presents the synthesis and characterization of lanthanum-modified alumina supported cerium-manganese mixed oxides, which were prepared by three different methods (coprecipitation, impregnation and citrate-based sol-gel method) followed by calcination at 500 °C. The physicochemical properties of the synthesized materials
Hydrothermal Synthesis of High Specific Capacity Al-doped V6O13 Cathode Material for Lithium-Ion Battery
Zhengguang Zou, et al.
International Journal of Electrochemical Science, 12, 1670-1679 (2017)
Ivan F Myronyuk et al.
Nanoscale research letters, 11(1), 153-153 (2016-03-24)
Transformation of Al(NO3)3∙9H2O (upon heating in the range of 20-1200 °C) into blends of amorphous and crystalline boehmite (210-525 °C), amorphous alumina and crystalline γ-Al2O3 (850 °C), and crystalline α-Al2O3 (1100 °C) was analyzed using X-ray diffraction (XRD), high-resolution transmission electron microscope (HRTEM), infrared
AlF 3-coated LiCoO 2 and Li [Ni 1/3 Co 1/3 Mn 1/3] O 2 blend composite cathode for lithium ion batteries.
Lee KS, et al.
Journal of Power Sources, 196(16), 6974-6977 (2011)
Fabrication of ?-alumina fibers by sol-gel and electrospinning of aluminum nitrate precursor solutions
Jose Hafid Roque-Ruiz, et al
Results in Physics, 12, 193-204 (2019)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service