289892
Gallium(III) nitrate hydrate
crystalline, 99.9% trace metals basis
Synonym(s):
Gallium trinitrate
About This Item
Recommended Products
Quality Level
Assay
99.9% trace metals basis
form
crystalline
reaction suitability
reagent type: catalyst
core: gallium
impurities
≤1500.0 ppm Trace Metal Analysis
mp
110 °C (dec.) (lit.)
solubility
H2O: slightly soluble(lit.)
ethanol: slightly soluble(lit.)
SMILES string
O.[Ga+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O
InChI
1S/Ga.3NO3.H2O/c;3*2-1(3)4;/h;;;;1H2/q+3;3*-1;
InChI key
YVFORYDECCQDAW-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Application
- Catalyst in the epoxidation of olefins in presence of hydrogen peroxide.
- Dopant in the preparation of ZnO nanorod arrays. The addition of Ga can improve the luminescence quality of ZnO nanorods.
- Precursor to synthesize Ga-MCM-41 materials (mesoporous silicate molecular sieves). Ga-MCM-41 can act as a catalyst in the liquid-phase isomerization of α-pinene.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2
Storage Class Code
5.1B - Oxidizing hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service