Skip to Content
Merck
All Photos(2)

Key Documents

325945

Sigma-Aldrich

Terbium(III) nitrate pentahydrate

99.9% trace metals basis

Synonym(s):

Terbium nitrate pentahydrate, Terbium(3+) nitrate pentahydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Tb(NO3)3 · 5H2O
CAS Number:
Molecular Weight:
435.02
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.9% trace metals basis

form

solid

reaction suitability

reagent type: catalyst
core: terbium

impurities

≤1500.0 ppm Trace Rare Earth Analysis

SMILES string

O.O.O.O.O.[Tb+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

InChI

1S/3NO3.5H2O.Tb/c3*2-1(3)4;;;;;;/h;;;5*1H2;/q3*-1;;;;;;+3

InChI key

YWROXJNVUWBEPC-UHFFFAOYSA-N

General description

Terbium(III) nitrate pentahydrate can be used as a dopant/precursor to prepare luminescent phosphors for various applications such as optical ceramics, white light emitting diodes, and photodynamic therapy.

Application

Terbium(III) nitrate pentahydrate can be used as:
  • A precursor to prepare Ce-doped terbium aluminum garnets by the photo-induced method.
  • A dopant to synthesize emission-tunable luminescent hydroxyapatite probe for bioimaging.
  • A starting material to prepare metal-organic frameworks for green laser applications.

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eulogio J Llorent-Martínez et al.
Analytical and bioanalytical chemistry, 410(2), 391-398 (2017-11-18)
We propose an alternative approach for the development of analytical methods based on terbium-sensitized luminescence (TSL). TSL is based on the complexation between Tb(III) ions and fluorescent organic compounds that have appropriate functional groups to complex with Tb(III). We report
I Perhaita et al.
Applied radiation and isotopes : including data, instrumentation and methods for use in agriculture, industry and medicine, 130, 188-197 (2017-10-06)
A series of novel apatite-based Ca
Reineke, T.M. et al.
Journal of the American Chemical Society, 121, 1651-1651 (1999)
Hassen Touzi et al.
Sensors (Basel, Switzerland), 21(5) (2021-03-07)
Gadolinium is extensively used in pharmaceuticals and is very toxic, so its sensitive detection is mandatory. This work presents the elaboration of a gadolinium chemical sensor based on 2-methylpyridine-substituted cyclam thin films, deposited on gold electrodes, using electrochemical impedance spectroscopy
Poonam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 152, 304-310 (2015-08-02)
The ternary terbium(III) complexes [Tb(HDAP)3⋅biq], [Tb(HDAP)3⋅dmph] and [Tb(HDAP)3⋅bathophen] were prepared by using methoxy substituted hydroxyketone ligand HDAP (2-hydroxy-4,6-dimethoxyacetophenone) and an ancillary ligand 2,2-biquinoline or 5,6-dimethyl-1,10-phenanthroline or bathophenanthroline respectively. The ligand and synthesized complexes were characterised based on elemental analysis, FT-IR

Articles

Innovation in dental restorative materials is driven by the need for biocompatible and natural-appearing restoration alternatives. Conventional dental materials like amalgam and composite resins have inherent disadvantages.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

Lanthanide ions in spectral conversion enhance solar cell efficiency via photon conversion.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service