All Photos(1)
About This Item
Linear Formula:
KO3SC6H4CO2H
CAS Number:
Molecular Weight:
240.27
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
95%
functional group
carboxylic acid
sulfonic acid
SMILES string
[K+].OC(=O)c1ccc(cc1)S([O-])(=O)=O
InChI
1S/C7H6O5S.K/c8-7(9)5-1-3-6(4-2-5)13(10,11)12;/h1-4H,(H,8,9)(H,10,11,12);/q;+1/p-1
InChI key
PXRJBUPXKDXDLG-UHFFFAOYSA-M
Related Categories
Application
4-Sulfobenzoic acid potassium salt was used as intercalating compound to improve clay exfoliation in the synthesis of poly(butylene terephthalate) (PBT) nanocomposites.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Berti C, et al.
Eur. Polymer J., 45(1), 70-78 (2009)
Pierre Thuéry et al.
Dalton transactions (Cambridge, England : 2003), 48(24), 8756-8772 (2019-05-24)
Nine uranyl ion complexes have been synthesized using two kinds of sulfonate-containing ligands, i.e. 2-, 3- and 4-sulfobenzoic acids (2-, 3- and 4-SBH2), which include additional carboxylic donors, and p-sulfonatocalix[4]arene (H8C4S), with additional phenolic groups, and [Ni(cyclam)]2+, [Cu(R,S-Me6cyclam)]2+ or PPh4+
Saccharin: para forms of some impurities are not mutagenic in Salmonella typhimurium.
F Poncelet et al.
Food and cosmetics toxicology, 18(4), 453-453 (1980-08-01)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service