Skip to Content
Merck
All Photos(4)

Key Documents

N9005

Sigma-Aldrich

1-Naphthylamine

97%

Synonym(s):

α-Naphthylamine, 1-Aminonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NH2
CAS Number:
Molecular Weight:
143.19
Beilstein:
386133
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

97%

form

solid

color

light tan to purple

bp

301 °C (lit.)

mp

47-50 °C (lit.)

density

1.114 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

Nc1cccc2ccccc12

InChI

1S/C10H9N/c11-10-7-3-5-8-4-1-2-6-9(8)10/h1-7H,11H2

InChI key

RUFPHBVGCFYCNW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

α-Naphthylamine, a naphthalene substituted by an amino group is an organic dye used as a detection reagent in the synthesis of novel azo dyes.[1]

Application

1-Naphthylamine has been used in the synthesis of Pitnot-2, an inactive analog of clathrin inhibitor Pitstop 2.[2]
It can also be used as a starting material to synthesize:
  • [(S)-HY-Phos], a novel chiral phosphine-phosphoramidite ligand for use in rhodium-catalyzed asymmetric hydrogenation of various functionalized olefins.[3]
  • N-(naphthalen-1-yl)picolinamide.[4]

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 2 - Carc. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synthesis of the Pitstop family of clathrin inhibitors.
Robertson M J, et al.
Nature Protocols, 9(7), 1592-1592 (2014)
Novel chiral phosphine-phosphoramidite ligands derived from 1-naphthylamine for highly efficient Rh-catalyzed asymmetric hydrogenation.
Yu S B, et al.
Tetrahedron Asymmetry, 19(15), 1862-1866 (2008)
Cobalt?Catalyzed, Aminoquinoline?Directed C (sp2)- H Bond Alkenylation by Alkynes.
Grigorjeva L and Daugulis O
Angewandte Chemie (International Edition in English), 126(38), 10373-10376 (2014)
Synthesis of dis-azo black dyes for electrowetting displays
Chiang Y, et al.
Journal of Materials Science and Engineering. B, 177, 1672-1677 (2012)
T C VandenBoer et al.
Journal of chromatography. A, 1618, 460843-460843 (2020-01-14)
A rapid separation and quantitation of the stereoisomer amino sugars glucosamine, galactosamine, and mannosamine, along with muramic acid, is needed. These compounds, when their quantities are accurate, can be used to understand the origin and fate of natural organic matter

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service