230901
4-Formylbenzene-1,3-disulfonic acid disodium salt hydrate
97%
Synonym(s):
Benzaldehyde 2,4-disulfonic acid disodium salt, Sodium 4-formylbenzene-1,3-disulfonate
About This Item
Recommended Products
Quality Level
Assay
97%
form
powder, crystals or chunks
application(s)
diagnostic assay manufacturing
hematology
histology
storage temp.
room temp
SMILES string
O.[Na+].[Na+].[O-]S(=O)(=O)c1ccc(C=O)c(c1)S([O-])(=O)=O
InChI
1S/C7H6O7S2.2Na.H2O/c8-4-5-1-2-6(15(9,10)11)3-7(5)16(12,13)14;;;/h1-4H,(H,9,10,11)(H,12,13,14);;;1H2/q;2*+1;/p-2
InChI key
YKJQVLWZGODFIH-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service